Skip to Content
Merck
All Photos(2)

Documents

CS0260

Sigma-Aldrich

Glutathione Assay Kit

sufficient for 700 assays

Synonym(s):

GSH Quantification Test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 700 assays

Quality Level

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... GSR(2936)

General description

The kit provides all reagents for simple and quick assay to measure the level of total glutathione (GSSG + GSH) in a cell and tissue extracts and in red blood cells or plasma. The sample is first deproteinized with the 5% 5-sulfosalicylic acid solution. Glutathione content of the sample is then assayed using a kinetic assay in which catalytic amounts of glutathione cause a continuous reduction of 5,5′-dithiobis-(2-nitrobenzoic) acid (DTNB) to TNB. The oxidised glutathione formed is recycled by glutathione reductase and NADPH. The product, TNB, is assayed colorimetrically at 412 nm.

Application

Glutathione Assay Kit has been used to measure total glutathione concentration in glutathione assay. It also has been used to determine different oxidative stress parameters in rats.

Biochem/physiol Actions

Glutathione (GSH) functions as a cofactor for several antioxidant enzymes. It is involved in the regeneration of vitamins C and E. It modulates cellular proliferation and apoptosis. GSH is crucial for mitochondrial functioning and the maintenance of mitochondrial DNA (mtDNA). Mutations in this gene are associated with Alzheimer′s, Parkinson′s and Huntington′s diseases. It is also linked to cystic fibrosis, immune diseases and cardiovascular diseases.
Reduced glutathione (GSH), a tripeptide (g-glutamyl-cysteinylglycine), is the major free thiol in most living cells and is involved in many biological processes such as detoxification of xenobiotics, removal of hydroperoxides, and maintenance of the oxidation state of protein sulfhydryls. It is the key antioxidant in animal tissues

Features and Benefits

  • A quick and simple method
  • The kit contains all the reagents required for a colorimetric assay of total glutathione (oxidised + reduced GSSG + GSH)
  • The kit includes a deproteination reagent for biological samples

Kit Components Only

Product No.
Description

  • Assay Buffer 5x 30 mL

  • DTNB 8 mg

  • DMSO 7.5 mL

  • Glutathine Reductase 20 U

  • L-Glutathione Reduced 300 μg

  • NADPH 25 mg

  • 5-Sulfosalicylic Acid 2.5 g

related product

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metallothionein-1G facilitates sorafenib resistance through inhibition of ferroptosis
Sun X, et al.
Hepatology, 64(2), 488-500 (2016)
Maria Swastika et al.
Malaria journal, 19(1), 208-208 (2020-06-20)
Glucose-6-phosphate dehydrogenase (G6PD) deficiency is the most common enzyme disorder in the world. Its main function is to generate NADPH that is required for anti-oxidative pathway in the cells especially in red blood cells (RBC). G6PD deficiency is X-linked and
Assay of glutathione, glutathione disulfide, and glutathione mixed disulfides in biological samples.
T P Akerboom et al.
Methods in enzymology, 77, 373-382 (1981-01-01)
Rat testicular impairment induced by electromagnetic radiation from a conventional cellular telephone and the protective effects of the antioxidants vitamins C and E
Al-Damegh MA
Clinics, 67(7), 785-792 (2012)
Genotoxicity of hydroquinone in A549 cells
Peng C, et al.
Cell Biology and Toxicology, 29(4), 213-227 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service