CLS431253
Corning® Erlenmeyer (Fernbach design) cell culture flasks
3 L Baffled Erlenmeyer (Fernbach Design) Flask w/ Vent Cap, polycarbonate, sterile, 4/cs
Synonym(s):
Corning Erlenmeyer flasks, cell culture flasks, conical flasks, plastic Erlenmeyer flasks, shaker flasks
About This Item
material
flat bottom flask
polycarbonate
polypropylene cap (vented)
vent cap
wide-mouth
wide-mouth neck
description
RNase- and DNase-free
sterility
sterile
feature
cap
disposable
Flask style: Erlenmeyer (Fernbach design)
packaging
pack of 1
case of 4
manufacturer/tradename
Corning 431253
flask capacity
3 L
color
orange cap
suitability
suitable for (Ideal for shaker culture applications)
Looking for similar products? Visit Product Comparison Guide
General description
- Fernbach design increases surface area for gas exchange
- Made from optically-clear polycarbonate
- Ideal for shaker culture applications
- Baffled bottoms improve mixing and increase gas exchange
- Vent caps supplied in every case of product for applications requiring sterile gas exchange
- Sterilized by gamma radiation and certified nonpyrogenic
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service