Skip to Content
MilliporeSigma

Skip To

CBA

Bile acids, conjugated

Synonym(s):

CBA, conjugated bile acids

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit

Available to ship TODAYfromMILWAUKEE

$249.00

About This Item

NACRES:
NA.26
UNSPSC Code:
12352202

$249.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

Biochem/physiol Actions

Unlike free bile acids, conjugated bile acids require an enzyme transporter for movement across the membrane of biliary epithelial cells.

Packaging

Conjugated Bile Acids in quantities indicated

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SMB00967B838148305
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Glycochenodeoxycholic Acid, Sodium Salt 25 mg
    SDS

  • Glycocholic Acid 100 mg
    SDS

  • Glycocholic Acid, Sodium Salt 100 mg
    SDS

  • Taurochenodeoxycholic Acid, Sodium Salt 25 mg
    SDS

  • Taurocholic Acid, Sodium Salt 250 mg
    SDS

  • Taurodeoxycholic Acid, Sodium Salt 1 g
    SDS

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters