Skip to Content
Merck

C8005

D-Cysteine hydrochloride monohydrate

≥98% (TLC)

Synonym(s):

D-Cysteine hydrochloride hydrate (1:1:1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HSCH2CH(NH2)COOH · HCl · H2O
CAS Number:
Molecular Weight:
175.63
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
NACRES:
NA.32
EC Number:
251-043-9
MDL number:
Beilstein/REAXYS Number:
5645478
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

D-Cysteine hydrochloride monohydrate, ≥98% (TLC)

Quality Segment

assay

≥98% (TLC)

form

powder or crystals

color

white to off-white

mp

185 °C

application(s)

peptide synthesis

SMILES string

Cl[H].[H]O[H].N[C@H](CS)C(O)=O

InChI

1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2/t2-;;/m1../s1

InChI key

QIJRTFXNRTXDIP-YBBRRFGFSA-N

Gene Information

Other Notes

Unnatural isomer of cysteine.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library