Skip to Content
Merck

Skip To

C7519

Choline bicarbonate

~80% in H2O

Synonym(s):

2-Hydroxyethyl)trimethylammonium bicarbonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C5H14NO · HCO3
CAS Number:
Molecular Weight:
165.19
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
50501514
MDL number:
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

concentration

~80% in H2O

density

1.17 g/mL at 25 °C (lit.)

lipid type

phospholipids

storage temp.

2-8°C

SMILES string

OC([O-])=O.C[N+](C)(C)CCO

InChI

1S/C5H14NO.CH2O3/c1-6(2,3)4-5-7;2-1(3)4/h7H,4-5H2,1-3H3;(H2,2,3,4)/q+1;/p-1

InChI key

DQKGOGJIOHUEGK-UHFFFAOYSA-M

General description

Choline acetyltransferase substrate

Application

Choline, a water soluble essential nutrient, is used by the body to make phosphatidylcholine phosphatides and neurotransmitter such as acetylcholine. Choline is used to study a wide range of biological processes from stem cell development to neurological disease studies. Also, An alkaline intermediate useful for preparing salts.

Biochem/physiol Actions

Acyl group acceptor

Other Notes

Note: The bicarbonate converts to the carbonate with release of CO2 on aging.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C16292697826978
form

liquid

form

powder

form

powder or crystals

form

powder or crystals

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

-

density

1.17 g/mL at 25 °C (lit.)

density

-

density

-

density

-

lipid type

phospholipids

lipid type

-

lipid type

-

lipid type

-

concentration

~80% in H2O

concentration

-

concentration

-

concentration

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
C7519-250ML04061837840593
C7519-100ML04061837840586
C7519-500ML04061837840609

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters