Skip to Content
Merck
All Photos(1)

Key Documents

C5743

Sigma-Aldrich

Cilastatin sodium salt

powder, ≥98% (HPLC)

Synonym(s):

Cilastatin sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H25N2NaO5S
CAS Number:
Molecular Weight:
380.43
EC Number:
UNSPSC Code:
12352204
NACRES:
NA.77

product name

Cilastatin sodium salt, ≥98% (HPLC)

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

powder

solubility

water: 10 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

InChI

1S/C16H26N2O5S.Na/c1-16(2)8-10(16)13(19)18-12(15(22)23)6-4-3-5-7-24-9-11(17)14(20)21;/h6,10-11H,3-5,7-9,17H2,1-2H3,(H,18,19)(H,20,21)(H,22,23);/q;+1/p-1/b12-6-;/t10-,11+;/m1./s1

InChI key

QXPBTTUOVWMPJN-QBNHLFMHSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cilastatin sodium salt is off-white to yellowish-white, hygroscopic, amorphous compound. It is derivatized from heptenoic acid. Its molecular mass is 380.43 g.

Biochem/physiol Actions

Inhibitor of the renal enzyme dehydropeptidase I and some bacterial zinc-containing β-lactamases. By these action, cilastatin suppresses both host and target metabolism of the broad-spectrum antibiotic imipenem, improving its efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinkyung Kim et al.
Proceedings of the National Academy of Sciences of the United States of America, 119(9) (2022-02-25)
Aminoglycosides (AGs) are commonly used antibiotics that cause deafness through the irreversible loss of cochlear sensory hair cells (HCs). How AGs enter the cochlea and then target HCs remains unresolved. Here, we performed time-lapse multicellular imaging of cochlea in live
First-order derivative ultraviolet spectrophotometry of imipenem-cilastatin formulations.
Tabbouche OS and Soukkariyyeh I
Journal of Taibah University Medical Sciences, 9(3), 178-181 (2014)
Saurav Roy Choudhury et al.
Cell, 178(5), 1205-1221 (2019-08-24)
A hallmark feature of inflammation is the orchestrated recruitment of neutrophils from the bloodstream into inflamed tissue. Although selectins and integrins mediate recruitment in many tissues, they have a minimal role in the lungs and liver. Exploiting an unbiased in vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service