Skip to Content
Merck
All Photos(1)

Key Documents

C3416

Sigma-Aldrich

Carbenicillin disodium salt

Carbenicillin disodium salt

BioReagent, suitable for plant cell culture

Synonym(s):

α-Carboxybenzylpenicillin disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2Na2O6S
CAS Number:
Molecular Weight:
422.36
Beilstein:
5722128
EC Number:
MDL number:
UNSPSC Code:
51282413
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

form

powder

technique(s)

cell culture | plant: suitable

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

agriculture

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)c3ccccc3)C(=O)N2[C@H]1C([O-])=O

InChI

1S/C17H18N2O6S.2Na/c1-17(2)11(16(24)25)19-13(21)10(14(19)26-17)18-12(20)9(15(22)23)8-6-4-3-5-7-8;;/h3-7,9-11,14H,1-2H3,(H,18,20)(H,22,23)(H,24,25);;/q;2*+1/p-2/t9?,10-,11+,14-;;/m1../s1

InChI key

RTYJTGSCYUUYAL-YCAHSCEMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Recommended for antibacterial use in cell culture media at 100 μg/ml and for selection of ampr transformed cells.

Biochem/physiol Actions

Mode of Action: Carboxypenicillin antibiotic that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane.


Antimicrobial spectrum: Active against Gram-positive and Gram-negative bacteria

Physical form

Hygroscopic powder

Analysis Note

Stable at 37 °C for 3 days

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Steinmann et al.
Euro surveillance : bulletin Europeen sur les maladies transmissibles = European communicable disease bulletin, 16(33) (2011-08-30)
We describe the epidemiology and characteristics of the pathogen and patients (n=7) associated with an outbreak of a carbapenem-resistant Klebsiella pneumoniae (CRKP) strain in a German university hospital from July 2010 to January 2011. Species identification and detection of carbapenem
Steven D Brown et al.
Biochemistry, 48(47), 11155-11157 (2009-10-24)
Bacteriophage Qbeta coat protein forms uniform virus-like particles when expressed recombinantly in a variety of organisms. We have inserted the IgG-binding Z domain at the carboxy terminus of the coat protein and coexpressed this chimeric subunit with native coat protein
Yasser M Abdelrahman et al.
Nucleic acids research, 39(5), 1843-1854 (2010-11-06)
Chlamydia trachomatis is an obligate intracellular bacterium that exhibits a unique biphasic developmental cycle that can be disrupted by growth in the presence of IFN-γ and β-lactams, giving rise to an abnormal growth state termed persistence. Here we have examined
Brigitte K Ziervogel et al.
Structure (London, England : 1993), 21(1), 76-87 (2012-12-04)
The structure of OmpF porin in complex with three common antibiotics (zwitterionic ampicillin, anionic ertapenem, and di-anionic carbenicillin) was determined using X-ray crystallography. The three antibiotics are found to bind within the extracellular and periplasmic pore vestibules, away from the
Catherine Kaminski et al.
Critical care (London, England), 15(2), R112-R112 (2011-04-13)
Although Pseudomonas aeruginosa is a leading pathogen responsible for ventilator-associated pneumonia (VAP), the excess in mortality associated with multi-resistance in patients with P. aeruginosa VAP (PA-VAP), taking into account confounders such as treatment adequacy and prior length of stay in

Articles

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Protocols

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service