Skip to Content
Merck
All Photos(4)

Key Documents

C2632

Sigma-Aldrich

CAPS

≥99%

Synonym(s):

3-(Cyclohexylamino)-1-propanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NH(CH2)3SO3H
CAS Number:
Molecular Weight:
221.32
Beilstein:
2835588
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99%

form

crystalline powder

pH

3.0-7.0

pKa (25 °C)

10.4

pKa 

10.4

mp

>300 °C (lit.)

solubility

H2O: soluble 15g + 85 mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

OS(=O)(=O)CCCNC1CCCCC1

InChI

1S/C9H19NO3S/c11-14(12,13)8-4-7-10-9-5-2-1-3-6-9/h9-10H,1-8H2,(H,11,12,13)

InChI key

PJWWRFATQTVXHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Frances E Lock et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(34), E3534-E3543 (2014-08-13)
Remnants of ancient transposable elements (TEs) are abundant in mammalian genomes. These sequences harbor multiple regulatory motifs and hence are capable of influencing expression of host genes. In response to environmental changes, TEs are known to be released from epigenetic
J H Jumppanen et al.
Electrophoresis, 16(8), 1441-1444 (1995-08-01)
In capillary zone electrophoresis the electrophoretic velocity of an ion decreases as the concentration of the background electrolyte solution is increased. This is caused by changes in the electrophoretic mobility of the ion (muep) as well as by changes in
Meng-Wei Hsieh et al.
Electrophoresis, 31(13), 2272-2278 (2010-07-02)
A dynamic reaction cell ICP-MS was used as a CE detector for the speciation analysis of arsenic and selenium. Samples containing arsenite, arsenate, monomethylarsonic acid, dimethylarsinic acid, arsenobetaine, selenite, selenate, selenocysteine, selenomethione, and Se-methylselenocysteine were subjected to electrophoretic separation before
Yi-Hsun Chang et al.
The Journal of biological chemistry, 282(47), 34306-34314 (2007-09-26)
3alpha-hydroxysteroid dehydrogenase/carbonyl reductase from Comamonas testosteroni catalyzes the oxidation of androsterone with NAD(+) to form androstanedione and NADH with a concomitant releasing of protons to bulk solvent. To probe the proton transfer during the enzyme reaction, we used mutagenesis, chemical
Els P M van Meijel et al.
BMC psychiatry, 15, 113-113 (2015-05-13)
Children and their parents are at risk of posttraumatic stress disorder (PTSD) following injury due to pediatric accidental trauma. Screening could help predict those at greatest risk and provide an opportunity for monitoring so that early intervention may be provided.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service