Skip to Content
MilliporeSigma

Skip To

C-22010

Endothelial Cell Growth Medium

Ready-to-use kit including Basal Medium and SupplementMix, 500 ml

Synonym(s):

Primary Cell Culture Media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL
Check Cart for Availability
$231.00

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207

$231.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Endothelial Cell Growth Medium (Ready-to-use)

Application

Endothelial Cell Growth Medium is a low-serum (2% V/V) medium optimized for the cultivation of endothelial cells from large blood vessels. The media are optimized for primary human cells but can also be used for bovine, porcine, murine, caprine, rabbit, elephant, dog, and rat endothelial cells from large vessels as well as for various endothelial cell lines.Note: For best transfection results we recommend using PromoCell heparin-free Endothelial Cell Growth Medium MV2. Heparin, included all our other our Endothelial Cell Growth Media formulations, may reduce transfection efficiency.

Packaging

Part of a Cell Culture System offered in a variety of formats:
Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix
Growth Medium Kit: Includes Basal Medium and SupplementPack
SupplementMix: Contains all media supplements premixed in one vial
SupplementPack: Contains all media supplements as individual vials

Preparation Note

Click here for more information.
Final supplement concentrations after addition to the basal medium:
Fetal Calf Serum: 0.02 ml / ml
Endothelial Cell Growth Supplement: 0.004 ml / ml
Epidermal Growth Factor (recombinant human): 0.1 ng / ml
Basic Fibroblast Growth Factor (recombinant human): 1 ng / ml
Heparin: 90 μg / ml
Hydrocortisone: 1 μg / ml
Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix or SupplementPack at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Analysis Note

All lots of PromoCell Endothelial Cell Media are subjected to comprehensive quality control tests using primary human endothelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested endothelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C-22110C-39210C-22210
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

liquid

form

liquid

form

liquid

form

aqueous solution

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

packaging

pkg of 500 ml

packaging

pkg of 500 ml

packaging

pkg of 1 kit

packaging

pkg of 500 ml


Still not finding the right product?

Explore all of our products under Endothelial Cell Growth Medium


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters