Skip to Content
MilliporeSigma

C1997

CYM-5442

≥98% (HPLC)

Synonym(s):

2-(4-(5-(3,4-diethoxyphenyl)-1,2,4-oxadiazol-3-yl)-2,3-dihydro-1H-inden-1-yl amino) ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H27N3O4
CAS Number:
Molecular Weight:
409.48
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

solubility

DMSO: >5 mg/mL

storage temp.

−20°C

SMILES string

CCOc1ccc(cc1OCC)-c2nc(no2)-c3cccc4C(CCc34)NCCO

InChI

1S/C23H27N3O4/c1-3-28-20-11-8-15(14-21(20)29-4-2)23-25-22(26-30-23)18-7-5-6-17-16(18)9-10-19(17)24-12-13-27/h5-8,11,14,19,24,27H,3-4,9-10,12-13H2,1-2H3

InChI key

NUIKTBLZSPQGCP-UHFFFAOYSA-N

Biochem/physiol Actions

CYM-5442 is a highly-selective S1P1 (Spingosine 1-Phosphate Receptor 1) agonist that activates S1P1-dependent pathways in vitro and to levels of full efficacy in vivo through a hydrophobic pocket, separable from the orthosteric site of S1P binding that is headgroup dependent. This is a different site from those involved in the activities of other agonists such as FTY720 and SEW2871.
CYM-5442 is a highly-selective S1P1 (Spingosine 1-Phosphate Receptor 1) agonist that activates S1P1-dependent pathways in vitro and to levels of full efficacy in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library