Skip to Content
Merck
All Photos(1)

Key Documents

B7559

Sigma-Aldrich

BW A4C

≥98% (HPLC)

Synonym(s):

N-[(E)-3-(3-Phenoxyphenyl)prop-2-enyl]acetohydroxamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17NO3
CAS Number:
Molecular Weight:
283.32
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white

mp

79.3-80.1 °C (lit.)

solubility

DMSO: 20 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)N(O)C\C=C\c1cccc(Oc2ccccc2)c1

InChI

1S/C17H17NO3/c1-14(19)18(20)12-6-8-15-7-5-11-17(13-15)21-16-9-3-2-4-10-16/h2-11,13,20H,12H2,1H3/b8-6+

InChI key

CEUDWZXMLMKPNN-SOFGYWHQSA-N

Gene Information

human ... ALOX5(240)
rat ... Alox5(25290)

Biochem/physiol Actions

Selective 5-lipoxygenase (5-LOX) inhibitor. Inhibits synthesis of LTB4.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S H Oliveira et al.
International archives of allergy and immunology, 111(3), 244-252 (1996-11-01)
Recently we demonstrated that the eosinophil migration into rat peritoneal cavities induced by a large volume of saline is mediated by LTB4 released by the resident macrophages and mast cells. In the present study, we have investigated the involvement of
Inhibition of leucotriene-synthesis improves dorsal skin flap survival in the rat.
L Törkvist et al.
Acta physiologica Scandinavica, 154(4), 511-512 (1995-08-01)
S A Yasin et al.
Journal of neuroendocrinology, 6(2), 179-184 (1994-04-01)
Interleukin-1 (IL-1) and interleukin-6 (IL-6) have been reported to stimulate the release of corticotrophin-releasing hormone (CRH) in vitro, the response being antagonized by the cyclo-oxygenase inhibitor, indomethacin. The effects of cytokines on the other major ACTH-releasing hormone, vasopressin (AVP), and
I D Brodowsky et al.
European journal of pharmacology, 254(1-2), 43-47 (1994-03-11)
Linoleic acid is converted to 8R-hydroperoxylinoleic acid by the soluble 8R-dioxygenase of the fungus Gaeumannomyces graminis. Effects of different lipoxygenase inhibitors on the 8R-dioxygenase were evaluated. Three hydroxamic acid derivatives were investigated. BW A4C (N-(3-phenoxycinnamyl)acetohydroxamic acid) was the most potent
C Zuany-Amorim et al.
British journal of pharmacology, 110(2), 917-924 (1993-10-01)
1. The intraperitoneal (i.p.) injection of 1 or 10 micrograms ovalbumin to sensitized Balb/c mice led to an acute histamine release, firstly evidenced 1 min after the challenge and returning to basal levels 30 min thereafter. This phenomenon was unaccompanied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service