Skip to Content
Merck

Skip To

B4532

5-Bromo-6-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt

≥98%

Synonym(s):

Magenta-GlcA, Magenta glucuronide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H13BrClNO7 · C6H13N
CAS Number:
Molecular Weight:
521.79
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

NC1CCCCC1.O[C@@H]2[C@@H](O)[C@@H](O[C@@H]([C@H]2O)C(O)=O)Oc3c[nH]c4cc(Cl)c(Br)cc34

InChI

1S/C14H13BrClNO7.C6H13N/c15-5-1-4-7(2-6(5)16)17-3-8(4)23-14-11(20)9(18)10(19)12(24-14)13(21)22;7-6-4-2-1-3-5-6/h1-3,9-12,14,17-20H,(H,21,22);6H,1-5,7H2/t9-,10-,11+,12-,14+;/m0./s1

InChI key

HGZDFBMYVMPFHR-CYRSAHDMSA-N

Application

Chromogenic substrate for β-glucuronidase; produces a magenta color in GUS+ bacterial colonies.

Other Notes

An alternative to 5-bromo-4-chloro-3-indolyl-β-D-glucuronide (X−GlcA) for detection of the β-glucuronidase gene.

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B5285B8049A4438
assay

≥98%

assay

≥98%

assay

-

assay

>98% (TLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

tablet

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
B4532-10MG04061833258705

Questions

Reviews

No rating value

Active Filters