Skip to Content
Merck
All Photos(2)

Documents

AV48268

Sigma-Aldrich

Anti-PRDX6 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-1-Cys, Anti-AOP2, Anti-AiPLA2, Anti-KIAA0106, Anti-MGC46173, Anti-NSGPx, Anti-PRX, Anti-Peroxiredoxin 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

25 kDa

species reactivity

horse, bovine, rat, rabbit, dog, human, mouse, guinea pig, sheep

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRDX6(9588)

General description

Peroxiredoxin 6 (PRDX6) is a thiol-specific antioxidant protein that acts as a bifunctional enzyme. It is involved in modulating the susceptibility of beta cells to oxidative stress. Prdx6 deficiency has been linked to impaired homeostasis and cell death/apoptosis. It has also been implicated in papillary thyroid carcinomas.
Rabbit Anti-PRDX6 antibody recognizes pig, bovine, human, mouse, rat, chicken, and zebrafish PRDX6.

Immunogen

Synthetic peptide directed towards the C terminal region of human PRDX6

Application

Rabbit Anti-PRDX6 antibody is suitable for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

PRDX6 is a member of the thiol-specific antioxidant protein family. This protein is a bifunctional enzyme with two distinct active sites. It is involved in redox regulation of the cell; it can reduce H(2)O(2) and short chain organic, fatty acid, and phospholipid hydroperoxides. It may play a role in the regulation of phospholipid turnover as well as in protection against oxidative injury. The protein encoded by this gene is a member of the thiol-specific antioxidant protein family. This protein is a bifunctional enzyme with two distinct active sites. It is involved in redox regulation of the cell; it can reduce H(2)O(2) and short chain organic, fatty acid, and phospholipid hydroperoxides. It may play a role in the regulation of phospholipid turnover as well as in protection against oxidative injury. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: VATPVDWKDGDSVMVLPTIPEEEAKKLFPKGVFTKELPSGKKYLRYTPQP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiqing Wang et al.
Neuroreport, 34(12), 606-616 (2023-07-03)
Subarachnoid hemorrhage (SAH) is an acute catastrophic neurological disorder with high morbidity and mortality. Ferroptosis is one of the pathophysiological processes during secondary brain injury of SAH, which could be inhibited by ferrostatin-1 (Fer-1) effectively. Peroxiredoxin6 (PRDX6) is an antioxidant
Arianna Nicolussi et al.
International journal of oncology, 44(2), 548-556 (2013-12-10)
Many clinical studies highlight the dichotomous role of PRDXs in human cancers, where they can exhibit strong tumor-suppressive or tumor-promoting functions. Recent evidence suggests that lower expression of PRDXs correlates with cancer progression in colorectal cancer (CRC) or in esophageal
Flavia M M Paula et al.
Molecular and cellular endocrinology, 374(1-2), 56-64 (2013-04-30)
Peroxiredoxins are a family of six antioxidant enzymes (PRDX1-6), and may be an alternative system for the pancreatic beta cells to cope with oxidative stress. This study investigated whether the main diabetogenic pro-inflammatory cytokines or the anti-inflammatory cytokine IL-4 modulate
Yanyun Jiang et al.
American journal of translational research, 9(5), 2374-2383 (2017-06-01)
Hyperglycemia-mediated damage to retinal pigment epithelial (RPE) cells plays a central role in the pathogenesis of diabetic retinopathy. Dysregulation of microRNA (miR)-383 modulates pancreatic beta cell survival in diabetes; however, its role in diabetic retinopathy remains unclear. In this study
Nigar Fatma et al.
American journal of physiology. Cell physiology, 301(4), C954-C967 (2011-06-17)
The multifunctional cytoprotective protein peroxiredoxin 6 (Prdx6) maintains cellular homeostasis and membrane integrity by regulating expression of intracellular reactive oxygen species (ROS) and phospholipid turnover. Using cells derived from targeted inactivation of Prdx6 gene or its depletion by RNA interference

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service