Skip to Content
Merck
All Photos(1)

Key Documents

AV39353

Sigma-Aldrich

Anti-DCP1A (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DCP1 decapping enzyme homolog A (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

rat, bovine, horse, guinea pig, human, rabbit, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DCP1A(55802)

General description

DCP1A is an mRNA decapping enzyme that is involved in TGF-β signaling. DCP1A is known to be hyper-phosphorylated during mitosis, cellular stress, neuronal differentiation and brain development. DCP1A has also been implicated in translational arrest.
Rabbit Anti-DCP1A antibody recognizes rat, human, chicken, bovine, canine, zebrafish, and mouse DCP1A.

Immunogen

Synthetic peptide directed towards the N terminal region of human DCP1A

Application

Rabbit Anti-DCP1A antibody is suitable for western blot applications at a concentration of 1.25 μg/ml.

Biochem/physiol Actions

Decapping is a key step in general and regulated mRNA decay. The protein encoded by this gene is a decapping enzyme. This protein and another decapping enzyme form a decapping complex, which interacts with the nonsense-mediated decay factor hUpf1 and may be recruited to mRNAs containing premature termination codons. This protein also participates in the TGF-beta signaling pathway.Decapping is a key step in general and regulated mRNA decay. The protein encoded by this gene is a decapping enzyme. This protein and another decapping enzyme form a decapping complex, which interacts with the nonsense-mediated decay factor hUpf1 and may be recruited to mRNAs containing premature termination codons. This protein also participates in the TGF-beta signaling pathway.

Sequence

Synthetic peptide located within the following region: MEALSRAGQEMSLAALKQHDPYITSIADLTGQVALYTFCPKANQWEKTDI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan D Dougherty et al.
The Journal of biological chemistry, 289(7), 3936-3949 (2014-01-03)
We have shown previously that poliovirus infection disrupts cytoplasmic P-bodies in infected mammalian cells. During the infectious cycle, poliovirus causes the directed cleavage of Dcp1a and Pan3, coincident with the dispersion of P-bodies. We now show that expression of Dcp1a
Adva Aizer et al.
PloS one, 8(1), e49783-e49783 (2013-01-10)
Processing bodies (PBs) are non-membranous cytoplasmic structures found in all eukaryotes. Many of their components such as the Dcp1 and Dcp2 proteins are highly conserved. Using live-cell imaging we found that PB structures disassembled as cells prepared for cell division
Jacob Blumenthal et al.
FEBS letters, 583(1), 197-201 (2008-12-17)
Decapping protein 1a (Dcp1a) is found in P-bodies and functions in mRNA cap removal prior to its degradation. The function and binding partners of Dcp1a have been thoroughly studied, however its expression pattern is still unclear. In this study we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service