Skip to Content
Merck

A6816

(±)-AMPA

solid

Synonym(s):

(±)-α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H10N2O4
Molecular Weight:
186.17
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Segment

color

white

solubility

H2O: 1 mg/mL (warm), 0.1 M HCl: 3.5 mg/mL, 0.1 M NaOH: 4.3 mg/mL, DMSO: 4.3 mg/mL, 1 M HCl: 50 mg/mL

SMILES string

O.Cc1onc(O)c1CC(N)C(O)=O

InChI

1S/C7H10N2O4.H2O/c1-3-4(6(10)9-13-3)2-5(8)7(11)12;/h5H,2,8H2,1H3,(H,9,10)(H,11,12);1H2

InChI key

HFISYNCCKQHIAM-UHFFFAOYSA-N

Application

(±)-AMPA has been used:
  • in vivo

  • whole-cell patch-clamp recording in Müller glial cells
  • to test its excitotoxic effects in hippocampal slice cultures
  • in retinal horizontal cells (HCs) culture to test its effect on Ca2+ signals

Biochem/physiol Actions

Potent excitatory amino acid that interacts selectively with central AMPA/kainate receptors.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library