Skip to Content
MilliporeSigma

Skip To

A273

Monoclonal Anti-Na+/K+ ATPase (α3 Subunit) antibody produced in mouse

1 mg/mL, clone XVIF9-G10, ascites fluid

Synonym(s):

Anti-AHC2, Anti-ATP1A1, Anti-CAPOS, Anti-DEE99, Anti-DYT12, Anti-RDP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.1 mg
Check Cart for Availability
$718.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
XVIF9-G10, monoclonal
Application:
IHC (f), WB
Citations:
5

$718.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

XVIF9-G10, monoclonal

mol wt

antigen ~110 kDa

species reactivity

human, rat, monkey, sheep, rabbit, guinea pig, canine

concentration

1 mg/mL

technique(s)

immunohistochemistry (frozen sections): 3 μg, western blot: 1 μg

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP1A3(478)
rat ... Atp1a3(24213)

General description

Mouse monoclonal anti-Na+/K+ ATPase (α3 subunit) antibody reacts specifically with Na+/K+ ATPase α3 subunit. By immunoblotting, the product reacts with human, monkey, sheep, canine, rabbit, guinea pig and rat tissues in which the α3 subunit is expressed. Immunohistochemical staining is consistent with the plasma membrane localization of Na+/K+ ATPase.
Na+/K+ transporting ATPase subunit α1 is an ion transport pump critical for maintaining the gradient of Na and K ions across the plasma membrane.

Immunogen

canine cardiac microsomes.

Application

Mouse monoclonal anti-Na+/K+ ATPase (α3 subunit) antibody has been used for chemiluminescent western blot analysis. The product has also been used for co-immunoprecipitation applications.

Physical form

Solution in phosphate buffered saline containing 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D219D218S1314
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

XVIF9-G10, monoclonal

clone

20A, monoclonal

clone

1A, monoclonal

clone

2A7-A1, monoclonal

Gene Information

human ... ATP1A3(478)
rat ... Atp1a3(24213)

Gene Information

mouse ... Cacna2d1(12293)
rat ... Cacna2d1(25399)

Gene Information

human ... CACNA1S(779)
mouse ... Cacna1s(12292)
rat ... Cacna1s(682930)

Gene Information

human ... ATP2A2(488)
mouse ... Atp2a2(11938)
rat ... Atp2a2(29693)

species reactivity

human, rat, monkey, sheep, rabbit, guinea pig, canine

species reactivity

rat, mouse, rabbit, guinea pig

species reactivity

human (weakly), guinea pig, mouse, rat, rabbit

species reactivity

guinea pig, human, canine, mouse, rat, rabbit

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

ascites fluid

biological source

mouse

biological source

mouse

biological source

-

biological source

mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters