Skip to Content
Merck
All Photos(1)

Documents

A2290

Sigma-Aldrich

Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

direct ELISA: 1:10,000
western blot: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders . Anti-Human IgG (γ-chain specific), (F(ab′)2) fragment-Peroxidase antibody is specific for human IgG when tested against human IgA, IgG, IgM, Bence Jones κ, and λ myeloma proteins.

Immunogen

Purified human IgG

Application

Anti-Human IgG (γ-chain specific), (F(ab′)2) fragment-Peroxidase antibody is suitable for use in ELISA and western blot .
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)
Peroxidase-conjugated goat anti-human IgG (Fab specific) antibody was used for western blot and ELISA analysis of yeast cultures genetically modified to produced recombinant Fab fragments. Anti-Human IgG antibody was used at a 1:10,000 dilution in PBS and incubated for 1 hour at room temperature. Binding in ELISA assays was detected with a O-phenylene- diamine substrate solution (Sigma).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4 containing 1% bovine serum albumin with preservative

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kiernan et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 8(9), 1829-1839 (2008-08-02)
Antibodies directed at non-gal xenoantigens are responsible for acute humoral xenograft rejection when gal knockout (GalTKO) pig organs are transplanted into nonhuman primates. We generated IgM and IgG gene libraries using peripheral blood lymphocytes of rhesus monkeys initiating active xenoantibody
Junzo Nojima et al.
Clinical chemistry, 51(3), 545-552 (2005-01-08)
Venous thromboembolic events such as deep vein thrombosis and pulmonary embolism are common manifestations of antiphospholipid syndrome. Our aim was to clarify the roles of anti-phospholipid (aPL) antibodies in the pathogenesis of venous thromboembolism (VTE) in patients with systemic lupus
Joanne L Zahorsky-Reeves et al.
BMC immunology, 7, 3-3 (2006-03-22)
The use of porcine cells and organs as a source of xenografts for human patients would vastly increase the donor pool; however, both humans and Old World primates vigorously reject pig tissues due to xenoantibodies that react with the polysaccharide
L D Ballam et al.
Journal of clinical pathology, 53(4), 314-317 (2000-05-24)
The salivary diagnosis of Helicobacter pylori infection offers attractive possibilities for the epidemiological study of infection in children. Salivary enzyme linked immunosorbent assay (ELISA) is less reliable then serum ELISA, owing to variable transudation of immunoglobulin. In addition, children are
Joanne L Zahorsky-Reeves et al.
Xenotransplantation, 14(2), 135-144 (2007-03-27)
Recent work has indicated a role for anti-Gal alpha 1-3Gal (Gal) and anti-non-Gal xenoantibodies in the primate humoral rejection response against human-decay accelerating factor (hDAF) transgenic pig organs. Our laboratory has shown that anti-porcine xenograft antibodies in humans and non-human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service