Skip to Content
Merck
All Photos(1)

Documents

A0862

Sigma-Aldrich

A-740003

≥98% (HPLC)

Synonym(s):

N-(1-{[(cyanoimino)(5-quinolinylamino) methyl] amino}-2,2-dimethylpropyl)-2-(3,4-dimethoxyphenyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30N6O3
CAS Number:
Molecular Weight:
474.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

COc1ccc(CC(=O)NC(N\C(Nc2cccc3ncccc23)=N\C#N)C(C)(C)C)cc1OC

InChI

1S/C26H30N6O3/c1-26(2,3)24(31-23(33)15-17-11-12-21(34-4)22(14-17)35-5)32-25(29-16-27)30-20-10-6-9-19-18(20)8-7-13-28-19/h6-14,24H,15H2,1-5H3,(H,31,33)(H2,29,30,32)

InChI key

PUHSRMSFDASMAE-UHFFFAOYSA-N

General description

A-740003 is a highly affine and selective P2X7 receptor antagonist and is best for the progression of a radiotracer for imaging of neuroinflammation by positron emission tomography.

Biochem/physiol Actions

A-740003 is a selective P2X7 purinoceptor antagonist.

Features and Benefits

This compound is featured on the P2 Receptors: P2X Ion Channel Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and initial preclinical evaluation of the P2X7 receptor antagonist [11C] A-740003 as a novel tracer of neuroinflammation
Janssen B, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 57(8), 509-516 (2014)
Pablo J Sáez et al.
PloS one, 8(8), e71361-e71361 (2013-08-27)
Reduced astrocytic gap junctional communication and enhanced hemichannel activity were recently shown to increase astroglial and neuronal vulnerability to neuroinflammation. Moreover, increasing evidence suggests that neuroinflammation plays a pivotal role in the development of Niemann-Pick type C (NPC) disease, an
Melissa Crabbé et al.
Frontiers in neuroscience, 13, 799-799 (2019-08-17)
The purinergic P2X7 receptor is a key mediator in (neuro)inflammation, a process that is associated with neurodegeneration and excitotoxicity in Parkinson's disease (PD). Recently, P2X7 imaging has become possible with [11C]JNJ-(54173)717. We investigated P2X7 availability, in comparison with availability of
Zihao Li et al.
Frontiers in cell and developmental biology, 9, 695041-695041 (2021-07-06)
Moderate-intensity exercise can help delay the development of osteoarthritis (OA). Previous studies have shown that the purinergic receptor P2X ligand gated ion channel 7 (P2X7) is involved in OA development and progression. To investigate the effect of exercise on P2X7

Articles

We offer many products related to P2X ion channel family receptors for your research needs.

We offer many products related to P2X ion channel family receptors for your research needs.

We offer many products related to P2X ion channel family receptors for your research needs.

We offer many products related to P2X ion channel family receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service