Skip to Content
MilliporeSigma

Skip To

A0230

Angiotensin IV trifluoroacetate salt

≥95% (HPLC), powder

Synonym(s):

Ang IV, Angiotensin II (3-8)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
$103.00

About This Item

Empirical Formula (Hill Notation):
C40H54N8O8 · xC2HF3O2
CAS Number:
Molecular Weight:
774.91 (free base basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥95% (HPLC)
Form:
powder

$103.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white

solubility

H2O: ~1 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](N)C(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](Cc4ccccc4)C(O)=O

InChI

1S/C40H54N8O8/c1-5-24(4)34(47-35(50)29(44-37(52)33(41)23(2)3)18-26-13-15-28(49)16-14-26)38(53)45-30(20-27-21-42-22-43-27)39(54)48-17-9-12-32(48)36(51)46-31(40(55)56)19-25-10-7-6-8-11-25/h6-8,10-11,13-16,21-24,29-34,49H,5,9,12,17-20,41H2,1-4H3,(H,42,43)(H,44,52)(H,45,53)(H,46,51)(H,47,50)(H,55,56)/t24-,29-,30-,31-,32-,33-,34-/m0/s1

InChI key

QSBGWDDCOJYQGY-KOQODJNWSA-N

Biochem/physiol Actions

AT4 angiotensin receptor agonist.
AT4 angiotensin receptor agonist. The AT4 receptor appears to be important in synaptic plasticity and thus in learning and memory.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML137410385SML0395
form

powder

form

powder

form

-

form

lyophilized solid

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPCE)

assay

≥95% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

H2O: ~1 mg/mL

solubility

-

solubility

H2O: 1.0 mg/mL, clear, colorless

solubility

-

color

white

color

white to beige

color

-

color

white


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters