Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
UNSPSC Code:
12352204
eCl@ss:
32160410
EC Number:
232-656-0
NACRES:
NA.54
MDL number:
Specific activity:
~8 U/mg
Biological source:
Canavalia ensiformis
biological source
Canavalia ensiformis
Quality Segment
form
powder
specific activity
~8 U/mg
mol wt
Mr ~480000
color
white
storage temp.
−20°C
General description
Subunit molecular weight: ~90,770
Composed of six subunits with total molecular weight: ~544,620
Composed of six subunits with total molecular weight: ~544,620
Urease (Ure) enzyme is present in the cytosol. It is active at acidic pH. Urease is present in archaea, bacteria, unicellular eukaryotes and plants.
Application
Urease from Canavalia ensiformis may be used for urea determination of various samples, such as legumes . It may be useful for the detection of pathogens as well as heavy-metal ions.
Biochem/physiol Actions
Urease catalyzes the hydrolysis of urea into carbon dioxide and ammonia. Urease is involved in nitrogen metabolism and urea degradation. Urease from Canavalia ensiformis binds 2 nickel ions per subunit .
Other Notes
1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol urea (Cat. No. 51459) per minute at pH 8.0 and 25°C
Still not finding the right product?
Explore all of our products under Urease from Canavalia ensiformis (Jack bean)
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 1
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

