Skip to Content
Merck

79470

D(+)-2-Phosphoglyceric acid sodium salt hydrate

≥75% (calc. on dry substance, enzymatic)

Synonym(s):

D-Glycerate 2-phosphate sodium salt, Sodium D-2-phosphoglycerate hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H7O7P · xNa+ · yH2O
CAS Number:
Molecular Weight:
186.06 (anhydrous free acid basis)
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.26
Assay:
≥75% (calc. on dry substance, enzymatic)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥75% (calc. on dry substance, enzymatic)

form

powder

impurities

≤1% Pi, ≤35% water

storage temp.

−20°C

SMILES string

O=C(O)[C@@H](CO)OP(O)(O)=O.C.C

InChI key

PXJAQZYAJDMQJH-YBBRRFGFSA-N

Biochem/physiol Actions

Enantiomerically pure metabolite of glycolysis/gluconeogenesis and also substrate for a number of important enzymes in central metabolism like enolase and phosphoglycerate mutase.
The enzyme enolase is responsible for reversible conversion of 2-phosphoglycerate (2-PG) into phosphoenolpyruvate (PEP). Phosphoglycerate mutase causes reversible isomerization of 3-phosphoglycerate and 2-phosphoglycerate in glycolysis and gluconeogenesis.


Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library