Skip to Content
Merck

68388

Phytic acid sodium salt hydrate

≥90% phosphorus (P) basis (dry basis)

Synonym(s):

myo-Inositol hexakis(dihydrogen phosphate), Inositol hexakisphosphate, Inositol hexaphosphoric acid, InsP6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H18O24P6 · xNa+ · yH2O
CAS Number:
Molecular Weight:
660.04 (anhydrous free acid basis)
UNSPSC Code:
12352201
MDL number:
NACRES:
NA.25
EC Number:
238-242-6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% phosphorus (P) basis (dry basis)

form

powder

impurities

≤0.1% inorganic phosphorus

loss

≤25.0% loss on drying

color

white

mp

53.27 -53.87  °C ((127.89 - 128.97 °F ) )

solubility

H2O: 50 mg/mL, clear to slightly hazy, colorless to light yellow

cation traces

Na: >10 (mole/m)

suitability

conforms to structure for Proton NMR spectrum

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)O[C@H]1[C@H](OP([O-])([O-])=O)[C@H](OP([O-])([O-])=O)[C@H](OP([O-])([O-])=O)[C@@H](OP([O-])([O-])=O)[C@@H]1OP([O-])([O-])=O

InChI

1S/C6H18O24P6.12Na/c7-31(8,9)25-1-2(26-32(10,11)12)4(28-34(16,17)18)6(30-36(22,23)24)5(29-35(19,20)21)3(1)27-33(13,14)15;;;;;;;;;;;;/h1-6H,(H2,7,8,9)(H2,10,11,12)(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24);;;;;;;;;;;;/q;12*+1/p-12

InChI key

KETSPIPODMGOEJ-UHFFFAOYSA-B

Application


  • Iron Fortification of Lentil (Lens culinaris Medik.) to Address Iron Deficiency.: This research explores the use of phytic acid sodium salt hydrate in iron fortification processes for lentils, addressing iron deficiency in human nutrition and enhancing the nutritional value of lentil-based diets (Podder et al., 2017).

Biochem/physiol Actions

Phytic acid is present in all eukaryotic cells. In plants, it is known to function as a [PO4]3− storage depot and precursor for other inositol phosphates and pyrophosphates. Its high surface negative charge makes it a potent chelator of divalent and trivalent cations in vitro, and it is most likely associated with Ca2+ or Mg2+ ions in vivo. In yeast, phytic acid acts with the nuclear pore protein Gle1 to regulate the nuclear export of mRNA by coactivating the RNA-dependent ATPase activity of DExD-box protein 5 (Dbp5).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?

Explore all of our products under Phytic acid sodium salt hydrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library