Skip to Content
Merck
All Photos(2)

Key Documents

402RA-05A

Sigma-Aldrich

Human Chondrocytes - Rheumatoid Arthritis: HC-RA, adult

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human articular cartilage (rheumatoid arthritic)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

chondrocyte

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

arthritis

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

HC-RA Human Chondrocytes are derived from human articular cartilage of donors with rheumatoid arthritis (RA). Rheumatoid arthritis is an inflammatory disease that is characterized by overproliferation of synovial cells and their infiltration into adjacent joint tissues, accompanied by gradual destruction of cartilage and bone erosion. Normally, chondrocytes are responsible for producing and maintaining the extracellular matrix of cartilage, but in RA cartilage, the environment is changed significantly due to inflammation and the invasion of other cells. Therefore, HC-RA can be a useful model to study the effects of those changes on chondrocyte biology. In parallel with normal HC, HC-RA can be used to investigate the differences between normal and pathological joints and evaluate potential treatment strategies.

Characterization: Positive for aggrecan after differentiation.

Cell Line Origin

Cartilage

Application

model to study effects of RA on chondrocytes, investigate differences between normal and pathological joints and evaluate potential treatment strategies

Components

Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 1st passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Subculture Routine

Please refer to the HC-RA Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service