Skip to Content
Merck
All Photos(1)

Key Documents

369A-1

Sigma-Aldrich

Phosphohistone H3 (PHH3) Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (369A-14)
vial of 0.5 mL concentrate (369A-15)
bottle of 1.0 mL predilute (369A-17)
vial of 1.0 mL concentrate (369A-16)
bottle of 7.0 mL predilute (369A-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... H3C1(8350)

General description

Phosphohistone H3 (PHH3) is a core histone protein, which together with other histones, forms the major protein constituents of the chromatin in eukaryotic cells. In mammalian cells, phosphohistone H3 is negligible during interphase but reaches a maximum for chromatin condensation during mitosis. Immunohistochemical studies showed anti-PHH3 specifically detected the core protein histone H3 only when phosphorylated at serine 10 or serine 28. Studies have also revealed no phosphorylation on the histone H3 during apoptosis. Therefore, PHH3 can serve as a mitotic marker to separate mitotic figures from apoptotic bodies and karyorrhectic debris, which may be a very useful tool in the diagnosis of tumor grades, especially in CNS, skin, gyn., soft tissue, and GIST.

Quality


IVD

IVD

IVD

RUO

Linkage

Phosphohistone H3 (PHH3) Positive Control Slides, Product No. 369S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoo-Jin Kim et al.
American journal of clinical pathology, 128(1), 118-125 (2007-06-21)
Mitotic activity is one of the most reliable prognostic factors in meningiomas. The identification of mitotic figures (MFs) and the areas of highest mitotic activity in H&E-stained slides is a tedious and subjective task. Therefore, we compared the results from
Howard Colman et al.
The American journal of surgical pathology, 30(5), 657-664 (2006-05-16)
Distinguishing between grade II and grade III diffuse astrocytomas is important both for prognosis and for treatment decision-making. However, current methods for distinguishing between grades based on proliferative potential are suboptimal, making identification of clear cutoffs difficult. In this study
Sayantani Goswami et al.
Journal of cellular physiology, 236(12), 8148-8159 (2021-07-01)
A number of studies have examined the effects of 1,25-dihydroxyvitamin D3 (1,25(OH)2 D3 ) on intestinal inflammation driven by immune cells, while little information is currently available about its impact on inflammation caused by intestinal epithelial cell (IEC) defects. Mice
M J Hendzel et al.
The Journal of biological chemistry, 273(38), 24470-24478 (1998-09-12)
Apoptosis plays an important role in the survival of an organism, and substantial work has been done to understand the signaling pathways that regulate this process. Characteristic changes in chromatin organization accompany apoptosis and are routinely used as markers for
Michel R Nasr et al.
The American Journal of dermatopathology, 30(2), 117-122 (2008-03-25)
Differentiating malignant melanoma from benign melanocytic lesions can be challenging. We undertook this study to evaluate the use of the immunohistochemical mitosis marker phospho-Histone H3 (pHH3) and the proliferation markers Ki-67 and survivin in separating malignant melanoma from benign nevi.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service