Skip to Content
MilliporeSigma

Skip To

31398

Dextran from Leuconostoc mesenteroides

Mr ~200,000

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
EC Number:
232-677-5
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(leuconostoc mesenteroides)

Quality Segment

form

powder

mol wt

Mr ~200,000

color

white

mp

483  °C ((901 °F ))

storage temp.

room temp

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Application

Dextran is a branched glucan composed of linear α(1→6) linked glucose units and α (1→3) link initiated branches. Dextran ranges in size from 10,000 to 150,000 Kd. Dextrans are used in many applications as volume extenders, stabilizers, matrix components, binding platforms, lubricants and physical structure components.
Use of dextrans as long and hydrophilic spacer arms improves the performance of immobilized proteins acting on macromolecules.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
313970918431389
biological source

(leuconostoc mesenteroides)

biological source

(Leuconostoc mesenteroides)

biological source

(Leuconostoc spp.)

biological source

(Leuconostoc spp)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

room temp

storage temp.

-

storage temp.

room temp

storage temp.

room temp

color

white

color

white

color

-

color

white

mp

483  °C ((901 °F ))

mp

483 °C ((901 °F ))

mp

483 °C ((901 °F))

mp

483  °C ((901 °F ))


Still not finding the right product?


Storage Class

11 - Combustible Solids

ppe

Eyeshields, Gloves, type N95 (US)

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters