14390C
Cellvento® 4CHO-C Cloning Medium
Animal- component free, chemically defined
About This Item
Recommended Products
sterility
sterile; sterile-filtered
product line
Cellvento®
form
liquid
technique(s)
cell culture | mammalian: suitable (suspension)
components
L-glutamine: no
shipped in
ambient
storage temp.
2-8°C
General description
Application
Features and Benefits
Packaging
Components
Caution
Reconstitution
Other Notes
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Cellvento® and EX-CELL® CHO cell culture media and feeds, optimized for monoclonal antibody & recombinant protein production in fed-batch & perfusion
Cellvento® and EX-CELL® CHO cell culture media and feeds, optimized for monoclonal antibody & recombinant protein production in fed-batch & perfusion
Related Content
Learn more about cell line development and the relevant products you need for this process.
Learn more about cell line development and the relevant products you need for this process.
Learn more about cell line development and the relevant products you need for this process.
Learn more about cell line development and the relevant products you need for this process.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service