Skip to Content
MilliporeSigma

Skip To

08599

N-Biotinyl-ethylenediamine trifluoroacetate salt

≥96.5% (HPLC)

Synonym(s):

N-(2-Aminoethyl)biotinamide trifluoroacetate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
$302.00
500 mg
Check Cart for Availability
$1,470.00
$1,029.00

About This Item

Empirical Formula (Hill Notation):
C12H22N4O2S · C2HF3O2
CAS Number:
Molecular Weight:
400.42
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Assay:
≥96.5% (HPLC)
Solubility:
DMSO: soluble, methanol: soluble

$302.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥96.5% (HPLC)

solubility

DMSO: soluble, methanol: soluble

storage temp.

2-8°C

InChI

1S/C12H22N4O2S.C2HF3O2/c13-5-6-14-10(17)4-2-1-3-9-11-8(7-19-9)15-12(18)16-11;3-2(4,5)1(6)7/h8-9,11H,1-7,13H2,(H,14,17)(H2,15,16,18);(H,6,7)/t8-,9-,11-;/m0./s1

InChI key

IVRQHQHWTOYIDN-GRIHUTHFSA-N

Application

Nucleophilic biotinylation reagent for the conjugation of various substrates and their identification and quantification using biotin-binding fluorescent labels

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
56951SML0013W3020
assay

≥96.5% (HPLC)

assay

≥95% (HPLC), ≥98% (T)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

solubility

DMSO: soluble, methanol: soluble

solubility

-

solubility

H2O: >2 mg/mL

solubility

H2O: >2 mg/mL

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters