T24007
Tetramethylsilane
ACS reagent, NMR grade, ≥99.9%
Synonym(s):
TMS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Si(CH3)4
CAS Number:
Molecular Weight:
88.22
Beilstein:
1696908
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
grade:
ACS reagent
NMR grade
NMR grade
Assay:
≥99.9%
technique(s):
NMR: suitable
bp:
26-28 °C (lit.)
vapor pressure:
11.66 psi ( 20 °C)
Recommended Products
grade
ACS reagent
NMR grade
Quality Level
vapor pressure
11.66 psi ( 20 °C)
Assay
≥99.9%
form
liquid
autoignition temp.
842 °F
expl. lim.
0.34-6.3 %
technique(s)
NMR: suitable
evapn. residue
≤0.05%
refractive index
n20/D 1.358 (lit.)
bp
26-28 °C (lit.)
mp
−99 °C (lit.)
density
0.648 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
C[Si](C)(C)C
InChI
1S/C4H12Si/c1-5(2,3)4/h1-4H3
InChI key
CZDYPVPMEAXLPK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Tetramethylsilane can be used as:
- A precursor to synthesize α-SiC by chemical vapor deposition.
- A reactant to synthesize 1,3,5,7-tetramethyladamantane from adamantane in the presence of aluminum chloride and dichloromethane.
- A solvent for the hydroaminomethylation of 1-octene with morpholine.
- A methylating reagent for the conversion of germanium tetrachloride to tetramethylgermanium, trimethylgermanium chloride, dimethylgermanium dichloride or methylgermanium trichloride.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-16.6 °F - closed cup
Flash Point(C)
-27 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tetramethylsilane in synthesis: selective mono-and polymethylations of germanium tetrachloride
Bordeau M, et al.
Organometallics, 1087-1089, 4-4 (1985)
A novel approach to selecting thermomorphic multicomponent solvent systems (TMS) for hydroaminomethylation reactions
Behr A, et al.
Chemical Engineering and Processing, 69, 15-23 (2013)
Synthesis of alpha-SiC from tetramethylsilane by chemical vapor deposition at high temperature
Jeong S, et al.
Applied Physics Express, 7, 025501-025501 (2014)
James C Salamanca et al.
Scientific reports, 8(1), 7559-7559 (2018-05-17)
E-cigarette aerosol emission studies typically focus on benchmarking toxicant levels versus those of cigarettes. However, such studies do not fully account for the distinct chemical makeup of e-liquids and their unique properties. These approaches often conclude that there are fewer
Exhaustive One-Step Bridgehead Methylation of Adamantane Derivatives with Tetramethylsilane
Bonsir M, et al.
European Journal of Organic Chemistry, 2021, 5227-5237 (2021)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service