Skip to Content
Merck
All Photos(4)

Key Documents

799319

Sigma-Aldrich

Aluminum oxide

activated, neutral, Brockmann I, free-flowing, Redi-Dri

Synonym(s):

Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
PubChem Substance ID:
NACRES:
NA.21

grade

Brockmann I
activated
standard grade

Quality Level

product line

Redi-Dri

form

powder

manufacturer/tradename

CAMAG 507-C-I

technique(s)

ion chromatography: suitable

surface area

155 m2/g

impurities

<0.03% Fe2O3
<0.03% SiO2
<0.4% Na2O

particle size

50-300 mesh

pore size

58 Å pore size

pH

7.0±0.5 ( in H2O)

mp

2040 °C (lit.)

anion traces

chloride (Cl-): ≤0.03 (mval/g)

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Brockmann aluminum oxides are suitable for column chromatography and adsorptive filtration.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon nanotube bundles self-assembled in double helix microstructures.
Cervantes-Sodi F, et al.
Carbon, 50(10), 3688-3693 (2012)
Prescott R and Graham MJ.
Oxidation of Metals, 38(3-4), 233-254 (1992)
Thermoanalytical studies of water on activated alumina, Brockmann I-V,(acid, neutral, basic) from- 60? to+ 700? C.
Hampson JW and Bleam WF.
Thermochimica Acta, 288(1), 179-189 (1996)
Colin J Ingham et al.
Biotechnology advances, 30(5), 1089-1099 (2011-08-23)
Porous aluminum oxide (PAO) is a ceramic formed by an anodization process of pure aluminum that enables the controllable assembly of exceptionally dense and regular nanopores in a planar membrane. As a consequence, PAO has a high porosity, nanopores with
Chien-Chih Lin et al.
Nanoscale, 5(17), 8090-8097 (2013-07-25)
We demonstrated a promising route for enhancing temperature sensitivity, improving saturation voltage, and reducing power consumption of the MOS(p) tunneling temperature sensors by introducing ultrathin Al2O3 into the dielectric stacks. Detailed illustrations of the working mechanism and device concept are

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service