Skip to Content
Merck
All Photos(3)

Key Documents

360554

Sigma-Aldrich

Nitromethane

ACS reagent, ≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3NO2
CAS Number:
Molecular Weight:
61.04
Beilstein:
1698205
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

2.1 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥95%

form

liquid

autoignition temp.

784 °F

expl. lim.

7.3 %, 33 °F

impurities

≤0.05% water

color

APHA: ≤10

refractive index

n20/D 1.382 (lit.)

pH

6.4 (20 °C, 0.01 g/L)

bp

101.2 °C (lit.)

mp

−29 °C (lit.)

density

1.127 g/mL at 25 °C (lit.)

suitability

clear for appearance

SMILES string

C[N+]([O-])=O

InChI

1S/CH3NO2/c1-2(3)4/h1H3

InChI key

LYGJENNIWJXYER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitromethane is an aliphatic nitro compound that is commonly used as a solvent for chemical processing and analysis.

Application

Nitromethane can be used as a solvent in the:
  • FeCl3-catalyzed Friedel-Crafts alkylation of indoles with alcohols.
  • Cobalt-catalyzed dehydration of aldoximes to nitriles.
  • Chemoselective oxidation of alcohols to carbonyl compounds.
  • Gold-catalyzed hydroarylation of aryl-substituted alkynes.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Metal ion encapsulation: cobalt cages derived from polyamines, formaldehyde, and nitromethane.
Geue RJ, et al.
Journal of the American Chemical Society, 106(19), 5478-5488 (1984)
The catalytic chemistry of nitromethane over Co-ZSM5 and other catalysts in connection with the methane-NOxSCR reaction.
Cowan AD, et al.
J. Catal., 176(2), 329-343 (1998)
Benedek Vakulya et al.
Organic letters, 7(10), 1967-1969 (2005-05-07)
Cinchona alkaloid-derived chiral bifunctional thiourea organocatalysts were synthesized and applied in the Michael addition between nitromethane and chalcones with high ee and chemical yields.
Ryan R Walvoord et al.
Organic letters, 14(16), 4086-4089 (2012-07-31)
An efficient cross-coupling reaction of aryl halides and nitromethane was developed with the use of parallel microscale experimentation. The arylnitromethane products are precursors for numerous useful synthetic products. An efficient method for their direct conversion to the corresponding oximes and
Huachang Hong et al.
The Science of the total environment, 444, 196-204 (2012-12-29)
The formations of THMs, HAAs, and HNMs from chlorination and chloramination of water from Jinlan Reservoir were investigated in this study. Results showed that monochloramine rather than chlorine generally resulted in lower concentration of DBPs, and the DBPs formation varied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service