Skip to Content
Merck
All Photos(3)

Key Documents

239232

Sigma-Aldrich

Calcium hydroxide

ACS reagent, ≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OH)2
CAS Number:
Molecular Weight:
74.09
EC Number:
MDL number:
UNSPSC Code:
12352305
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥95.0%
grade:
ACS reagent
form:
powder

grade

ACS reagent

Quality Level

Assay

≥95.0%

form

powder

impurities

≤0.03% insol. in HCl
≤0.1% S compounds
≤3.0% calcium carbonate

pH

12.4-12.6 (20 °C)

density

2.24 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.03%

cation traces

Fe: ≤0.05%
K: ≤0.05%
Mg: ≤0.5%
Na: ≤0.05%
Sr: ≤0.05%
heavy metals: ≤0.003% (by ICP)

SMILES string

O[Ca]O

InChI

1S/Ca.2H2O/h;2*1H2/q+2;;/p-2

InChI key

AXCZMVOFGPJBDE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium hydroxide is commonly used in wastewater treatment plants (WWTPs) to adjust pH. Calcium hydroxide has numerous applications due to its strong basic characteristics. Uses include flocculants in water and sewage treatment, a whitewash ingredient, soil improvement, as a lye substitute in non-lye hair relaxers, and in the liming industry to neutralize the acid.

Application

Calcium hydroxide can be used:
  • In the pretreatment of technical lignin during the pyrolysis in a fluidized bed reactor to overcome the agglomeration to produce phenolic-rich bio-oil.
  • As a catalyst in the pyrolysis of pinewood in a fixed bed reactor.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The antibacterial effect of camphorated paramonochlorophenol, camphorated phenol and calcium hydroxide in the treatment of infected root canals.
A Bystrom et al.
Endodontics & dental traumatology, 1(5), 170-175 (1985-10-01)
U Schröder
Journal of dental research, 64 Spec No, 541-548 (1985-04-01)
The findings from the recent literature on pulpal cell responses to the application of calcium hydroxide to exposed pulps are described. The effect of calcium hydroxide on healthy and inflamed pulp is discussed. The effect of incorporation of calcium hydroxide
Lisbet Mailin López González et al.
Bioresource technology, 131, 250-257 (2013-01-29)
Different pre-treatment severities by thermo-alkaline conditions (100°C, Ca(OH)2) on press mud were evaluated for different pre-treatment time and lime loading. COD solubilization and the methane yield enhancement were assessed. The biochemical methane potential was determined in batch assays under mesophilic
Nikte Gómez-Ortíz et al.
ACS applied materials & interfaces, 5(5), 1556-1565 (2013-01-26)
The presence and deteriorating action of microbial biofilms on historic stone buildings have received considerable attention in the past few years. Among microorganisms, fungi are one of the most damaging groups. In the present work, antimicrobial surfaces were prepared using
J F Siqueira et al.
International endodontic journal, 32(5), 361-369 (1999-11-07)
Literature review The primary function of calcium hydroxide as a routine intracanal medicament is to provide antimicrobial activity. However, the mechanisms of antimicrobial activity of calcium hydroxide are not well known. Physicochemical properties of this substance may limit its effectiveness

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service