Skip to Content
Merck
All Photos(4)

Key Documents

216003

Sigma-Aldrich

Ammonium phosphate monobasic

ACS reagent, ≥98%

Synonym(s):

APM, mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

vapor pressure

<0.001 hPa ( 20 °C)

Assay

≥98%

form

crystals or chunks

impurities

≤0.005% NH4OH pptd.
≤0.005% insolubles

pH

3.8-4.4 (25 °C, 5%)

mp

190 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 ppm
nitrate (NO3-): ≤0.001%
sulfate (SO42-): ≤0.01%

cation traces

Ca: 0.001%
Fe: ≤0.001%
K: ≤0.005%
Mg: 0.005%
Na: ≤0.005%
heavy metals (as Pb): ≤5 ppm

SMILES string

N.OP(O)(O)=O

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the synthesis of the first example of a mixed-valence ammonium iron phosphate, NH4Fe2(PO4)2.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milne GWA.
Gardner's Commercially Important Chemicals: Synonyms, Trade Names, and Properties, 193-193 (2005)
Burdock GA.
Encyclopedia of Food and Color Additives, 142-142 (1997)
Boudin, S. Lii, K-H.
Inorganic Chemistry, 37, 799-799 (1998)
Maria Lina Sanna et al.
Fungal biology, 116(7), 769-777 (2012-07-04)
Pichia fermentans DiSAABA 726 is a biofilm-forming yeast that undergoes dimorphic transition. Under yeast-like morphology it controls brown rot caused by Monilia spp. on apple fruit, while under pseudohyphal form, it shows pathogenic behaviour itself on peach fruit. The present
Everest Akaraonye et al.
Biotechnology journal, 7(2), 293-303 (2011-12-08)
The main hindrance in the use of polyhydroxyalkanoates (PHAs) as a replacement for existing petroleum-based plastics is their high production cost. The carbon source accounts for 50% of the cost for PHA production. Thus, increasing the yield and productivity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service