Skip to Content
Merck
All Photos(1)

Key Documents

Y0001393

Lincomycin hydrochloride for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Lincomycin hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34N2O6S · HCl · H2O
CAS Number:
Molecular Weight:
461.01
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lincomycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

Cl[H].[H]O[H].CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O

InChI

1S/C18H34N2O6S.ClH.H2O/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4;;/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25);1H;1H2/t9-,10-,11+,12-,13+,14-,15-,16-,18-;;/m1../s1

InChI key

LFZGYTBWUHCAKF-DCNJEFSFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lincomycin hydrochloride for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reza Tabrizi et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 72(5), 850-857 (2014-02-25)
The aim of the present study was to evaluate the periodontal regenerative capacity of demineralized freeze-dried bone allograft (DFDBA) alone or used with local lincomycin. In the present single-blind, randomized, controlled clinical trial, 20 subjects 26 years old or older
Qunfei Zhao et al.
Nature, 518(7537), 115-119 (2015-01-22)
Low-molecular-mass thiols in organisms are well known for their redox-relevant role in protection against various endogenous and exogenous stresses. In eukaryotes and Gram-negative bacteria, the primary thiol is glutathione (GSH), a cysteinyl-containing tripeptide. In contrast, mycothiol (MSH), a cysteinyl pseudo-disaccharide
Biochemistry: Elusive source of sulfur unravelled.
Charles E Melançon
Nature, 518(7537), 45-46 (2015-01-22)
Milena Marsoni et al.
Ecotoxicology and environmental safety, 108, 52-57 (2014-07-22)
Pharmaceutically active compounds (PACs) are continuously dispersed into the environment due to human and veterinary use, giving rise to their potential accumulation in edible plants. In this study, Eruca sativa L. and Zea mays L. were selected to determine the
Leslie C Dickson
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 967, 203-210 (2014-08-16)
This paper describes an extension and performance characterization of a quantitative confirmatory multi-residue liquid chromatography-tandem mass spectrometric method for residues of macrolide and lincosamide antibiotics, originally validated for application to bovine kidney tissues, to tissues of salmon, shrimp and tilapia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service