Skip to Content
Merck
All Photos(1)

Documents

Y0001298

Chlorambucil for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Chlorambucil, 4-(4-[Bis(2-chloroethyl)amino]phenyl)butyric acid, 4-[Bis(2-chloroethyl)amino]benzenebutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19Cl2NO2
CAS Number:
Molecular Weight:
304.21
Beilstein:
999011
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

chlorambucil

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CCCc1ccc(cc1)N(CCCl)CCCl

InChI

1S/C14H19Cl2NO2/c15-8-10-17(11-9-16)13-6-4-12(5-7-13)2-1-3-14(18)19/h4-7H,1-3,8-11H2,(H,18,19)

InChI key

JCKYGMPEJWAADB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlorambucil for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R G Palmer et al.
Cancer treatment reviews, 11(2), 121-129 (1984-06-01)
There is a high incidence of acute myeloid leukaemia in patients who have been treated with chlorambucil. This appears at least in part to be directly due to the drug itself and cannot be attributed to the disease processes for
A Begleiter et al.
Leukemia & lymphoma, 23(3-4), 187-201 (1996-10-01)
Chronic lymphocytic leukemia (CLL) is the most common leukemia in Western countries but the clinical presentation and rate of disease progression are highly variable. When treatment is required the most commonly used therapy is the nitrogen mustard alkylating agent, chlorambucil
R A Luqmani et al.
Bailliere's clinical rheumatology, 4(3), 595-619 (1990-12-01)
Immunosuppressive agents serve a major role in the management of once-fatal conditions such as the systemic necrotizing vasculitides, but they are also being used in more common, chronic inflammatory disorders such as rheumatoid arthritis. The drugs are all capable of
S Machado et al.
European journal of dermatology : EJD, 11(5), 458-462 (2001-08-30)
We report a case of necrobiotic xanthogranuloma in a 51 year-old white male patient presenting with a 6-year history of multiple indurated violaceous nodules and plaques involving the eyelids, trunk and extremities. He had an associated paraproteinemia (Ig G lambda)
E M Rinchik et al.
BioEssays : news and reviews in molecular, cellular and developmental biology, 15(12), 831-836 (1993-12-01)
Recent mutagenesis studies have demonstrated that the chemotherapeutic agent, chlorambucil (CHL), is highly mutagenic in male germ cells of the mouse. Post-meiotic germ cells, and especially early spermatids, are the most sensitive to the cytotoxic and mutagenic effects of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service