Skip to Content
Merck
All Photos(1)

Documents

PHR1513

Supelco

Enrofloxacin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Enrofloxacin, ENR, ENRO, Baytril

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22FN3O3
CAS Number:
Molecular Weight:
359.39
Beilstein:
5307824
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001259
traceable to USP 1235900

API family

enrofloxacin

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN1CCN(CC1)c2cc3N(C=C(C(O)=O)C(=O)c3cc2F)C4CC4

InChI

1S/C19H22FN3O3/c1-2-21-5-7-22(8-6-21)17-10-16-13(9-15(17)20)18(24)14(19(25)26)11-23(16)12-3-4-12/h9-12H,2-8H2,1H3,(H,25,26)

InChI key

SPFYMRJSYKOXGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Enrofloxacin is classified under the second generation quinolone antibiotics, known as ′fluoroquinolones′. It is a component of several medicines used to treat urinary tract infections, infections of the gastrointestinal tract, respiratory tract infections, infections of the skin structure as well as surgical prophylaxis.

Application

Enrofloxacin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and water samples by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0027 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of a novel HPLC sorbent material for the determination of ten quinolones in human and veterinary pharmaceutical formulations.
Samanidou VF, et al.
Journal of Separation Science, 28(18), 2444-2453 (2005)
Multi-class determination of around 50 pharmaceuticals, including 26 antibiotics, in environmental and wastewater samples by ultra-high performance liquid chromatography?tandem mass spectrometry.
Gracia-Lor E, et al.
Journal of Chromatography A, 1218(16), 2264-2275 (2011)
Withdrawal time of four pharmaceutical formulations of enrofloxacin in poultry according to different maximum residues limits.
Martin SB, et al.
Journal of Veterinary Pharmacology and Therapeutics, 33(3), 246-251 (2010)
Enrofloxacin assay validation and pharmacokinetics following a single oral dose in chickens.
Da Silva RG, et al.
Journal of Veterinary Pharmacology and Therapeutics, 29(5), 365-372 (2006)
Taylor B Engle et al.
BMC veterinary research, 10, 286-286 (2014-12-05)
Porcine circovirus 2 is the primary agent responsible for inducing a group of associated diseases known as Porcine Circovirus Associated Diseases (PCVAD), which can have detrimental effects on production efficiency as well as causing significant mortality. The objective of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service