Skip to Content
Merck
All Photos(1)

Documents

PHR1499

Supelco

Flurbiprofen

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Flurbiprofen, (±)-2-Fluoro-α-methyl-4-biphenylacetic acid, L-790,330

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H3(F)CH(CH3)CO2H
CAS Number:
Molecular Weight:
244.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. F0285200
traceable to USP 1285750

API family

flurbiprofen

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

110-112 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC(C(O)=O)c1ccc(c(F)c1)-c2ccccc2

InChI

1S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10H,1H3,(H,17,18)

InChI key

SYTBZMRGLBWNTM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Flurbiprofen is a non-selective, non-steroidal anti-inflammatory drug (NSAID) that is widely used against rheumatoid arthritis. It is also effective against vernal keratoconjunctivitis, post-operative ocular inflammation, herpetic stromal keratitis, excimer laser photorefractive keratectomy and ocular gingivitis.

Application

Flurbiprofen may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and biological samples by various analytical techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Fluibiprofen is a cyclooxygenase (COX) inhibitor, which is an enzyme responsible for the conversion of arachidonic acid to prostaglandin G2 (PGG2) and PGG2 to prostaglandin H2 (PGH2) in the prostaglandin synthesis pathway. This decreases the prostaglandins which cause inflammation, pain, swelling and fever. Flurbiprofen inhibits the activity of both COX-1 and -2. The S enantiomer inhibits prostaglandin synthesis and has both anti-inflammatory and analgesic activity, while the R enantiomer does not inhibit prostaglandin synthesis and displays only analgesic activity.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3047 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The determination of non-steroidal antiinflammatory drugs in pharmaceuticals by capillary zone electrophoresis and micellar electrokinetic capillary chromatography.
Donato MG, et al.
Journal of Pharmaceutical and Biomedical Analysis, 12(1), 21-26 (1994)
Determination of flurbiprofen in pharmaceutical formulations by UV spectrophotometry and liquid chromatography.
Sajeev C, et al.
Analytica Chimica Acta, 463(2), 207-217 (2002)
Sample pretreatment and determination of non steroidal anti-inflammatory drugs (NSAIDs) in pharmaceutical formulations and biological samples (blood, plasma, erythrocytes) by HPLC-UV-MS and μ-HPLC.
Mohamed S, et al.
Current Medicinal Chemistry, 12(5), 573-588 (2005)
Simultaneous Determination of Ceftriaxone Sodium and Non Steroidal Anti?Inflammatory Drugs in Pharmaceutical Formulations and Human Serum by RP?HPLC.
Sultana N, et al.
J. Chin. Chem. Soc., 57(6), 1278-1285 (2010)
Philippe Denis et al.
Investigative ophthalmology & visual science, 56(2), 1089-1096 (2015-01-22)
To evaluate the safety and efficacy of high-intensity focused ultrasound (HIFU) cyclocoagulation in reducing intraocular pressure (IOP) in patients with refractory glaucoma by using a novel miniaturized delivery device (EyeOP1). We conducted a 12-month open-label multicenter prospective study (EyeMUST1 Study).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service