Skip to Content
Merck
All Photos(2)

Key Documents

M8277

Supelco

Midodrine hydrochloride

analytical standard, for drug analysis

Synonym(s):

(±)-1-(2,5-Dimethoxyphenyl)-2-glycinamidoethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O4 · HCl
CAS Number:
Molecular Weight:
290.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.COc1ccc(OC)c(c1)C(O)CNC(=O)CN

InChI

1S/C12H18N2O4.ClH/c1-17-8-3-4-11(18-2)9(5-8)10(15)7-14-12(16)6-13;/h3-5,10,15H,6-7,13H2,1-2H3,(H,14,16);1H

InChI key

MGCQZNBCJBRZDT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Midodrine is a prodrug and a long acting α-adrenergic stimulating agent, and forms an active metabolite, desglymidodrine, which causes an elevation of the systemic blood pressure with the reduction in heart rate.

Application

Midodrine hydrochloride may be used as an analytical reference standard for the quantification of the analyte in biological samples and pharmaceutical formulations using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

α-Adrenergic agonist; vasoconstrictor

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda B Hassinger et al.
Pediatric nephrology (Berlin, Germany), 28(2), 345-348 (2012-09-21)
Denys-Drash (DDS) syndrome is a rare genetic syndrome resulting from a mutation in the Wilms' tumor suppressor gene 1 (WT1), which presents with early onset nephrotic syndrome progressing rapidly to end-stage kidney disease (ESKD), pseudohermaphroditism, and high rates of Wilms'
Douglas Z Liou et al.
Chest, 144(3), 1055-1057 (2013-09-07)
Thoracic duct injury is a rare but serious complication following surgery of the neck or chest that leads to uncontrolled chyle leak. Conventional management includes drainage, nutritional modification, or aggressive surgical interventions such as thoracic duct ligation, flap coverage, fibrin
Khurram Bari et al.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association, 10(10), 1169-1175 (2012-07-18)
Large-volume paracentesis (LVP) is the treatment of choice for patients with cirrhosis and refractory ascites. However, LVP can lead to postparacentesis circulatory dysfunction (PCD), which is associated with faster ascites recurrence and renal failure. PCD results from vasodilatation, which reduces
Amanda J Ross et al.
Clinical science (London, England : 1979), 126(4), 289-296 (2013-08-28)
POTS (postural tachycardia syndrome) is a chronic form of OI (orthostatic intolerance). Neuropathic POTS is characterized by decreased adrenergic vasoconstriction, whereas hyperadrenergic POTS exhibits increased adrenergic vasoconstriction. We hypothesized that midodrine, an α1-adrenergic receptor agonist, would increase CVR (calf vascular
Determination of midodrine in human plasma by high-performance liquid chromatography with fluorescence detection
Yoshida H, et al.
Analytical Sciences, 19(2), 317-319 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service