Skip to Content
MilliporeSigma

FPPMCC2

Flash Point Standard - Pensky Martens

certified reference material, ASTM D 93 Proc. A (76.5 °C)

Synonym(s):

Pensky Martens Flash Point Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

according to ASTM® D93

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

petroleum
pharmaceutical

format

neat

General description

Flash Point Standards- Pensky Martens is a certified reference material that is tested and certified in accordance with ASTM D93 ′Standard Test Method for Flash Point by Pensky Martens Closed Cup Tester - Procedure A′. Procedure A is applicable to distillate fuels - diesels, biodiesel blends, kerosene, heating oil, turbine fuels and lubricating oils. Typically used in, but not limited to, the analysis of petroleum products with flash points above 40 °C and below 360 °C.
Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Pensky Martens Flash Point Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials


Still not finding the right product?


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

168.8 °F

flash_point_c

76 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library