Skip to Content
Merck
All Photos(1)

Key Documents

D2954600

Docosahexaenoic acid ethyl ester

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4,7,10,13,16,19-Docosahexaenoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36O2
CAS Number:
Molecular Weight:
356.54
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

docosahexaenoic acid ethyl ester

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O(CC)C(=O)CC\C=C\C\C=C\C\C=C\C\C=C\C\C=C\C\C=C\CC

InChI

1S/C24H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26-4-2/h5-6,8-9,11-12,14-15,17-18,20-21H,3-4,7,10,13,16,19,22-23H2,1-2H3/b6-5+,9-8+,12-11+,15-14+,18-17+,21-20+

InChI key

ITNKVODZACVXDS-YATCGRJWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Docosahexaenoic acid ethyl ester EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Tang et al.
Lipids in health and disease, 11, 70-70 (2012-06-09)
Bioactivities of Docosahexaenoic acid (DHA) and Eicosapentaenoic acid (EPA) depend on their chemical forms. The present study was to investigate short term effects of triglyceride (TG), ethyl ester (EE), free fatty acid (FFA) and phospholipid (PL) forms of omega-3 fatty
Hidefumi Yoshii et al.
Bioscience, biotechnology, and biochemistry, 66(4), 749-753 (2002-05-31)
The application of omega-3 polyunsaturated fatty acids (PUFAs) as food additives is restricted by their chemically quite reactive properties. However, quantitative analyses of the oxidative kinetics of PUFAs are very few compared to other studies on food chemistry. In this
P Green et al.
Biochimica et biophysica acta, 1531(1-2), 156-164 (2001-03-30)
Previously we have shown that intraamniotic administration of ethyl docosahexaenoate (Et-DHA) to pregnant rats resulted in decreased lipid peroxidation in the fetal brain, under a variety of conditions (S. Glozman, P. Green, E. Yavin, J. Neurochem. 70 (1998) 2482-2491). In
Alfonso Valenzuela et al.
Annals of nutrition & metabolism, 49(1), 49-53 (2005-03-01)
Docosahexaenoic acid (C22:6, DHA) is an omega-3 fatty acid required for the normal development of the mammalian nervous and visual system. DHA is provided by the mother during pregnancy and lactating period. Mother's DHA supplementation during pregnancy, and even before
K Umegaki et al.
Free radical research, 34(4), 427-435 (2001-05-01)
We compared the influence of docosahexaenoic acid (DHA) supplementation on oxidative DNA damage in bone marrow between young and aged rats. As a marker of oxidative DNA damage, 8-hydroxydeoxyguanosine (8-OHdG) in DNA was analyzed. Young (5-week-old) and aged (100-week-old) female

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service