Skip to Content
Merck
All Photos(1)

Key Documents

BCR048R

Benzo[k]fluoranthene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1873745
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

215-217 °C (lit.)

solubility

95% ethanol: <1 mg/mL at 20 °C
DMSO: <1 mg/mL at 20 °C
H2O: <1 mg/mL at 20 °C
acetone: 1-10 mg/mL at 20 °C
methanol: <1 mg/mL at 20 °C
toluene: 5-10 mg/mL at 20 °C

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2cc-3c(cc2c1)-c4cccc5cccc-3c45

InChI

1S/C20H12/c1-2-6-15-12-19-17-10-4-8-13-7-3-9-16(20(13)17)18(19)11-14(15)5-1/h1-12H

InChI key

HAXBIWFMXWRORI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR048R

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Q Pan et al.
Environmental pollution (Barking, Essex : 1987), 141(3), 443-451 (2005-11-08)
The effects of benzo(a)pyrene (BaP), benzo(k)fluoranthene (BkF) and their mixture on antioxidant enzyme activities and lipid peroxidation (LPO) levels of haemolymph of scallop (Chlamys ferrari) were studied. The superoxide dismutase (SOD) activities of 0.5 microg/L and 1.0 microg/L were significantly
Erin E Bessette et al.
Drug metabolism and disposition: the biological fate of chemicals, 33(3), 312-320 (2004-12-04)
Polycyclic aromatic hydrocarbons (PAHs) and heavy metals are often environmental cocontaminants that could interact to alter PAH carcinogenicity. The heavy metal, arsenite, and the PAH, benzo[k]fluoranthene, were used as prototypes to investigate, in human HepG2 cells, mechanisms whereby the bioactivation
G Verrhiest et al.
Ecotoxicology (London, England), 10(6), 363-372 (2002-01-05)
Polycyclic Aromatic Hydrocarbons (PAHs) are ubiquitous pollutants of sediments. Sediment quality criteria often use toxicity data for individual PAHs. However, PAHs always occur in field sediments as a complex mixture of compounds. In this study, the toxicity of phenanthrene (P)
Tohru Saitoh et al.
Talanta, 79(2), 177-182 (2009-06-30)
A simple and rapid method for the highly sensitive determination of polycyclic aromatic hydrocarbons (PAHs) in water was developed. Benzo[a]pyrene, benzo[k]fluoranthene, perylene, and pyrene in water were concentrated into sodium dodecyl sulfate (SDS)-alumina admicelles. The collection was performed by adding
Hiroyuki Masaki et al.
The Analyst, 130(9), 1253-1257 (2005-08-13)
We developed a device and some systems for detecting benzo[a]pyrene (B[a]P) and benzo[k]fluoranthene (B[k]F). The device uses a UV light-emitting diode that emits light with a wavelength of 370 nm and a violet laser diode that emits light with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service