Skip to Content
Merck
All Photos(1)

Key Documents

95410

Sigma-Aldrich

Tungsten(VI) oxide

powder, puriss., 99.9%

Synonym(s):

Tungstic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WO3
CAS Number:
Molecular Weight:
231.84
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

grade

puriss.

Quality Level

Assay

99.9%

form

powder

density

7.16 g/mL at 25 °C (lit.)

SMILES string

O=[W](=O)=O

InChI

1S/3O.W

InChI key

ZNOKGRXACCSDPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tungsten(VI) oxide (WO3) is a semiconducting material with excellent electrochromic properties. It is a cost efficient material with a high optical modulation. It can be synthesized by a variety of methods which include chemical vapor deposition, electrodeposition, and other sol-gel based coating processes.

Application

WO3 can be used for a wide range of applications such as electrochemical devices, photocatalysis, biological sensors, and other display devices.

Caution

may contain dark blue particles.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamalakannan Kailasam et al.
ChemSusChem, 8(8), 1404-1410 (2015-03-25)
Composites of mesoporous polymeric carbon nitride and tungsten(VI) oxide show very high photocatalytic activity for the evolution of hydrogen from water under visible light and in the presence of sacrificial electron donors. Already addition of very small amounts of WO3
KARAKTERISTIK GEL TITANIUM TUNGSTAT DAN PENGARUHNYA TERHADAP PELEPASAN RENIUM-188
Setiawan D, et al.
Jurnal Iptek Nuklir Ganendra, 16(1), 1-8 (2013)
Low-firing thick-film piezoresistive sensors for medical instruments
Maeder T, et al.
Sensors and actuators A, Physical, 172(1), 228-232 (2011)
Inkjet printing of sol-gel derived tungsten oxide inks
Vidmar T, et al.
Solar Energy Materials and Solar Cells, 125, 87-95 (2014)
Electrochemical reduction of tungsten compounds to produce tungsten powder
Erdogan M and Karakaya I
Metall. Mater. Trans. B (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service