Skip to Content
Merck
All Photos(1)

Documents

75285

Supelco

N′-Nitrosonornicotine

analytical standard

Synonym(s):

1-Nitroso-2-(3-pyridyl)pyrrolidine, 3-(1-Nitroso-2-pyrrolidinyl)pyridine, NNN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N3O
CAS Number:
Molecular Weight:
177.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O=NN1CCCC1c2cccnc2

InChI

1S/C9H11N3O/c13-11-12-6-2-4-9(12)8-3-1-5-10-7-8/h1,3,5,7,9H,2,4,6H2

InChI key

XKABJYQDMJTNGQ-UHFFFAOYSA-N

General description

N′-Nitrosonornicotine is a tobacco specific nitrosamine, which is found to be carcinogenic in nature. It is produced via nitrosation of nicotine during the process of curing, aging, processing and smoking of tobacco.

Application

N′-Nitrosonornicotine may be used as an analytical reference standard for the quantification of the analyte in biological samples using liquid chromatography coupled with tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Besson et al.
PloS one, 12(6), e0178795-e0178795 (2017-06-09)
For marine fishes with a bipartite life cycle, pelagic larval dispersal can shape the distribution, connectivity, composition and resilience of adult populations. Numerous studies of larval dispersal, and associated settlement and recruitment processes, have examined the relationship between population connectivity
Comparative carcinogenicity in A/J mice and metabolism by cultured mouse peripheral lung of N'-nitrosonornicotine, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone, and their analogues
Castonguay A, et al.
Cancer Research, 43(3), 1223-1229 (1983)
Bastian Niemeyer et al.
Molecular ecology resources, 17(6), e46-e62 (2017-05-11)
Reliable information on past and present vegetation is important to project future changes, especially for rapidly transitioning areas such as the boreal treeline. To study past vegetation, pollen analysis is common, while current vegetation is usually assessed by field surveys.
Julia Heger-Stevic et al.
PLoS pathogens, 14(12), e1007488-e1007488 (2018-12-20)
Hepatitis B virus (HBV) replicates its 3 kb DNA genome through capsid-internal reverse transcription, initiated by assembly of 120 core protein (HBc) dimers around a complex of viral pregenomic (pg) RNA and polymerase. Following synthesis of relaxed circular (RC) DNA
Nafiseh Ghobakhloo et al.
Tropical medicine & international health : TM & IH, 24(1), 116-126 (2018-02-16)
Leishmania major has been considered as the main aetiological agent of cutaneous leishmaniasis in Iran. However, there are recent reports about the existence of Crithidia spp in cutaneous lesions in southern Iran. Therefore, this study was designed to decipher some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service