Skip to Content
Merck
All Photos(2)

Key Documents

74553

Supelco

Rubiadin

analytical standard

Synonym(s):

1,3-Dihydroxy-2-methyl-9,10-anthracenedione, 1,3-Dihydroxy-2-methylanthraquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O4
CAS Number:
Molecular Weight:
254.24
Colour Index Number:
75350
Beilstein:
1885558
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

O=C1C2=C(C=C(O)C(C)=C2O)C(C3=CC=CC=C31)=O

InChI

1S/C15H10O4/c1-7-11(16)6-10-12(13(7)17)15(19)9-5-3-2-4-8(9)14(10)18/h2-6,16-17H,1H3

InChI key

IRZTUXPRIUZXMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L R Comini et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(12), 1093-1095 (2011-06-15)
Searching for agents that could be effective in the treatment of cancer, special highlight has focused on the study of numerous plant-derived compounds. We previously demonstrated that anthraquinones (AQs) isolated from a vegetal species: Heterophyllaea pustulata Hook f. (Rubiaceae), such
Y Kawasaki et al.
Chemical & pharmaceutical bulletin, 40(6), 1504-1509 (1992-06-01)
Twenty compounds were isolated from the roots of Rubia tinctorum which are used as a commercial source of madder color. Among these compounds, mollugin (1), 1-hydroxy-2-methylanthraquinone (2), 2-ethoxymethylanthraquinone(11), rubiadin (13), 1,3-dihydroxyanthraqunone (14), 7-hydroxy-2-methylanthraquinone (16), lucidin (17), 1-methoxymethylanthraquinone (18) and lucidin-3-O-primeveroside
Susana C Núñez Montoya et al.
Toxicon : official journal of the International Society on Toxinology, 51(8), 1409-1415 (2008-06-03)
Heterophyllaea pustulata (Rubiaceae), a South American genus, is a phototoxic shrub that grows in the Andean mountain range of the northwest of Argentina, popularly known as "cegadera". Animals that ingest the aerial parts of this plant suffer a typical primary
Y B Tripathi et al.
Indian journal of biochemistry & biophysics, 35(5), 313-316 (1999-07-20)
The inhibition of FeSO4 induced lipid peroxidation in rat liver by alcoholic extract of Rubia cordifolia and by one of its constituent rubiadin (1, 3-dihydroxy-2-methyl anthraquinone) (pure form) has been compared. Both have been found to inhibit lipid peroxidation in
Wei-Hua Huang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 33(5), 524-526 (2008-06-10)
To investigate the chemical constituents from Hedyotis diffusa. The compounds were isolated and purified by various chromatographic techniques and identified by their physicochemical properties and spectral data. Eight compounds were isolated and identified as octadecyl (E)-p-coumarate (1), p-E-methoxy-cinnamic acid (2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service