Skip to Content
Merck
All Photos(1)

Documents

71662

Sigma-Aldrich

di-Sodium hydrogen phosphate dihydrate

tested according to Ph. Eur.

Synonym(s):

Dinatrii phosphas dihydricus, sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 2H2O
CAS Number:
Molecular Weight:
177.99
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

Assay

98.0-101.0% (dried substance)

form

solid

SMILES string

O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.2H2O/c;;1-5(2,3)4;;/h;;(H3,1,2,3,4);2*1H2/q2*+1;;;/p-2

InChI key

KDQPSPMLNJTZAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium phosphate dibasic dihydrate has been used in buffer preparation for tandem purification of His-FLAG tagged protein.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tandem purification of His6-3x FLAG tagged proteins for mass spectrometry from Arabidopsis
Huang He and Nusinow D
Bio-protocol, 6, e2060-e2060 (2016)
Christopher Polonyi et al.
Dalton transactions (Cambridge, England : 2003), 43(31), 11941-11949 (2014-06-27)
The ability of the anticancer active drug cisplatin to exert biological activity through interference with nucleic acid function is well documented. Since kinetics play a key role in determining product distributions in these systems, methods for accurate documentation of reactivity
Susann K J Ludwig et al.
Analytical and bioanalytical chemistry, 406(27), 6857-6866 (2014-06-29)
Current contaminant and residue monitoring throughout the food chain is based on sampling, transport, administration, and analysis in specialized control laboratories. This is a highly inefficient and costly process since typically more than 99% of the samples are found to
M J Hilhorst et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 965, 45-53 (2014-07-06)
A liquid chromatography-tandem mass spectrometry method is described for the quantitative determination of artemether (ART) and its metabolite dihydroartimisinin (DHA) in human plasma samples. Quantitation of ART and DHA in plasma is challenging due to the presence of malaria related
M O Besenhard et al.
International journal of pharmaceutics, 475(1-2), 198-207 (2014-08-12)
We present a proof-of-concept study of a continuous coating process of single API crystals in a tubular reactor using coacervation as a microencapsulation technique. Continuous API crystal coating can have several advantages, as in a single step (following crystallization) individual

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service