Skip to Content
Merck
All Photos(1)

Key Documents

71300

Sigma-Aldrich

Sodium benzoate

purum p.a., ≥99.0% (NT)

Synonym(s):

Benzoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COONa
CAS Number:
Molecular Weight:
144.10
Beilstein:
3572467
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39024301
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Quality Level

Assay

≥99.0% (NT)

form

powder

mp

>300 °C (lit.)

solubility

water: soluble(lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤2000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

WXMKPNITSTVMEF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium benzoate is the sodium salt of benzoic acid. On dissolution in water it affords weakly basic solution. It is widely employed as a food preservative and in various cosmetic products. It has anti-corrosive properties. Its determination in fruit juices, sodas, soy sauce, ketchup, peanut butter, cream cheese and other foods by HPLC method has been proposed. It exhibits microbicidal action and is added as anti-microbial agent in various food products. It is a Food and Drug Administration-approved nontoxic drug.

Application

Sodium benzoate may be used in the preparation of various heavy metal benzoates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saurav Brahmachari et al.
Journal of immunology (Baltimore, Md. : 1950), 179(1), 275-283 (2007-06-21)
Experimental allergic encephalomyelitis (EAE) is the animal model for multiple sclerosis. This study explores a novel use of sodium benzoate (NaB), a commonly used food additive and a Food and Drug Administration-approved nontoxic drug for urea cycle disorders, in treating
Eagleson M.
Concise Encyclopedia Chemistry, 121-121 (1994)
Andrew G Reidenbach et al.
Biomolecules, 10(1) (2019-12-22)
Antisense oligonucleotides (ASOs) designed to lower prion protein (PrP) expression in the brain through RNase H1-mediated degradation of PrP RNA are in development as prion disease therapeutics. ASOs were previously reported to sequence-independently interact with PrP and inhibit prion accumulation
H M Pylypiw et al.
Journal of chromatography. A, 883(1-2), 299-304 (2000-07-26)
A rapid and reliable method is presented for the determination of the preservatives sodium benzoate and potassium sorbate in fruit juices, sodas, soy sauce, ketchup, peanut butter, cream cheese, and other foods. The procedure utilizes high-performance liquid chromatography (HPLC) followed
Huey-Jen Tsay et al.
Neurotoxicology and teratology, 29(5), 562-569 (2007-07-24)
Sodium benzoate (SB) is a commonly used food preservative and anti-microbial agent in many foods from soup to cereals. However, little is known about the SB-induced toxicity and teratogenicity during early embryonic development. Here, we used zebrafish as a model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service