Skip to Content
Merck
All Photos(1)

Documents

68281

Supelco

Alkane standard mixture for performance tests of GC-systems

analytical standard, C10 - C40 (all even), 50 mg/l each

Synonym(s):

Determination of hydrocarbons (even-numbered) according to ISO 16703 and EN 14039

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EN 14039
ISO 16703

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.388

density

0.684 g/mL at 20 °C

application(s)

petroleum

format

multi-component solution

Application

Alkane standard mixture has been used as a retention index calibration standard in the metabolic analysis of body fluids of humans and vertebrates using gas chromatography coupled with mass spectrometry (GC-MS).
It may also be used for performance tests of GC-systems as per DIN ISO 9377-2:2000 method used for the determination of petroleum hydrocarbons in water samples by gas chromatography technique.
Alkane standard mixture has been used as a retention index calibration standard in the metabolic analysis of body fluids of humans and vertebrates using gas chromatography coupled with mass spectrometry(GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Components

contains all even n-alkanes from C10 to C40, 50 mg/L in n-heptane each

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cristian Varela et al.
Scientific reports, 10(1), 14716-14716 (2020-09-09)
Over the course of human history and in most societies, fermented beverages have had a unique economic and cultural importance. Before the arrival of the first Europeans in Australia, Aboriginal people reportedly produced several fermented drinks including mangaitch from flowering
Metabolic profiling of body fluids and multivariate data analysis
Trezzi P-J, et al.
MethodsX, 4, 95-103 (2017)
Katie C Hootman et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(21), E4233-E4240 (2017-05-10)
Metabolomic markers associated with incident central adiposity gain were investigated in young adults. In a 9-mo prospective study of university freshmen (
Challenges in the determination of petroleum hydrocarbons in water by gas chromatography (hydrocarbon index)
Drozdova.S, et al.
Fuel: The Science and Technology of Fuel and Energy, 113, 527-536 (2013)
Lisa Krämer et al.
Metabolites, 8(1) (2018-02-15)
Currently, changes in metabolic fluxes following consumption of stable isotope-enriched foods are usually limited to the analysis of postprandial kinetics of glucose. Kinetic information on a larger diversity of metabolites is often lacking, mainly due to the marginal percentage of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service