Skip to Content
Merck
All Photos(1)

Key Documents

66006BC

Millipore

SimPlate® Campylobacter CI

BioControl, Detection and quantification of Campylobacter coli and Campylobacter jejuni in poultry rinses and meat

Synonym(s):

Campy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
material:
plastic vial (containing dehydrated culture media)
suitability:
Campylobacter spp.
feature:
fluorescent indicator

material

plastic vial (containing dehydrated culture media)

size

100 tests

form

dry powder

shelf life

6‑18 mo.

feature

fluorescent indicator

packaging

container of 100

manufacturer/tradename

BioControl Systems 66006-100

storage condition

dry media

application(s)

food and beverages

compatibility

for use with SimPlate (65009BC)

storage temp.

room temp

suitability

Campylobacter spp.

General description

SimPlate® Campylobacter CI is designed for the detection and quantification of Campylobacter coli and Campylobacter jejuni. SimPlate® Binary Detection Technology combines proprietary media and a patented plating device to provide fast and accurate quantitative results for key groups of indicator organisms. Defined substrates incorporated into the media are utilized to detect multiple enzymes and redox reactions associated with the target organisms, which results in chromogenic or fluorescent changes to the media, and provides easy-to-read results. Because SimPlate® relies on these key reactions rather than the formation of a colony, results are available days faster than conventional culture methods.

Application

SimPlate® Campylobacter CI is used in the detection and quantification of Campylobacter coli and Campylobacter jejuni in poultry rinses and meat samples. SimPlate® tests have been validated by international standards (AOAC/MicroVal)

Features and Benefits

  • Proprietary chromogenic media and patented plating device with isolation wells combine to produce easily counted results in just 48 hours.
  • While the traditional agar plate reference method requires the use of 4 Campy CEFEX agar plates to analyze 1 mL sample, SimPlate Campylobacter has been validated to provide equivalent results with just a single plate. Combined with the reduction of necessary sample dilutions, made possible by the broad counting range of the plate (1-738), SimPlate reduces number of tests required for each sample, helping to reduce overall testing costs.

Components

1 package of 50 single-test vials

Testing Method

Binary Detection Technology®

Other Notes

Required for use: SimPlate Device (65009BC)

Legal Information

Binary Detection Technology is a registered trademark of Merck KGaA, Darmstadt, Germany
SimPlate is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • SimPlate Campylobacter Single Dose, Irradiated

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service