Skip to Content
Merck
All Photos(1)

Key Documents

51161

Millipore

Pseudomonas ID Membrane

suitable for microbiology, white colored, sterile, diameter 70 mm

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

membrane size

70 mm

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Pseudomonas spp.

General description

Pseudomonas ID Membrane is a substrate-carrying membrane used for rapid identification and differentiation of Pseudomonas through characteristic chromogenic reactions while placed on the agar surface post-incubation. The membranes contain chromogenic substrates, which serve as the basis for microbe differentiation by color. The target organisms possess enzyme systems that metabolize the substrates in the membrane, leading to a change of color. The colors can be visually detected on the membrane or under UV at 365 nm.

Application

Pseudomonas ID Membrane is used for rapid detection & confirmation of Pseudomonas aeruginosa from clinical and nonclinical specimens. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service