Skip to Content
Merck
All Photos(1)

Documents

50725

Millipore

Nocive Brewers Bacteria Broth Base, modified

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Detection Broth for bacterial beer spoilers, Pedi-Lacto Selective Beer Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

beef extract, 2 g/L
chlorophenol red, 0.07 g/L
L-cysteine hydrochloride, 0.2 g/L
dextrose, 15 g/L
disodium phosphate, 2 g/L
L-malic acid, 0.5 g/L
maltose, 15 g/L
pancreatic digest of casein, 5 g/L
polysorbate 80, 0.5 g/L
potassium acetate, 6 g/L
yeast extract, 5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

5.8±0.2 (25 °C)

application(s)

bioburden testing
food and beverages
sample preparation

microbiology

suitability

Leuconostoc spp.
Pediococcus spp.
selective for Lactobacillus spp.
selective for Lactococcus spp.

Application

Selective medium used for the detection of contaminating/spoilage microorganisms in brewery.

Preparation Note

Suspend 51,3 grams of the medium in 500 ml of distilled water and 500 ml of beer without gas. Mix well. Heat for one minute with frequent agitation until the medium is completely dissolved. Sterilize in autoclave at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

HybriScan™ assays provide rapid, accurate molecular detection of beer spoilage microorganisms, ensuring excellent product quality in breweries.

HybriScan™ assays provide rapid, accurate molecular detection of beer spoilage microorganisms, ensuring excellent product quality in breweries.

HybriScan™ assays provide rapid, accurate molecular detection of beer spoilage microorganisms, ensuring excellent product quality in breweries.

HybriScan™ assays provide rapid, accurate molecular detection of beer spoilage microorganisms, ensuring excellent product quality in breweries.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service