Skip to Content
Merck
All Photos(1)

Documents

46502

Supelco

Nitrofurantoin

VETRANAL®, analytical standard

Synonym(s):

N-(5-Nitro-2-furfurylidene)-1-aminohydantoin, Furadoxyl, Nitrofurantoine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N4O5
CAS Number:
Molecular Weight:
238.16
Beilstein:
893207
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

cleaning products
clinical
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

Mode of action

DNA synthesis | interferes
cell wall synthesis | interferes
protein synthesis | interferes

SMILES string

[O-][N+](=O)c1ccc(\C=N\N2CC(=O)NC2=O)o1

InChI

1S/C8H6N4O5/c13-6-4-11(8(14)10-6)9-3-5-1-2-7(17-5)12(15)16/h1-3H,4H2,(H,10,13,14)/b9-3+

InChI key

NXFQHRVNIOXGAQ-YCRREMRBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitrofurantoin is an antibiotic which is effective against both gram-positive and gram-negative bacteria. The drug is primarily used to treat urinary tract infections.

Application

Nitrofurantoin has been used as a reference standard for the determination of nitrofurantoin in milk by high-performance liquid chromatography with electrochemical detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of nitrofurantoin in pharmaceutical preparations using flow injection-spectrophotometry.
Had H, et al.
Journal of the Association of Arab Universities for Basic and Applied Sciences, 24(1), 74-80 (2017)
Hedvig Nordeng et al.
Obstetrics and gynecology, 121(2 Pt 1), 306-313 (2013-01-25)
To estimate whether first-trimester exposure to nitrofurantoin is associated with increased malformation rates and, secondarily, to estimate whether exposure to nitrofurantoin is associated with increased rates of negative pregnancy outcomes and perinatal complications. We conducted a population-based cohort study using
Benjamin A Rogers et al.
Microbial drug resistance (Larchmont, N.Y.), 19(2), 100-103 (2013-01-22)
The New Delhi metallo-β-lactamase gene (bla(NDM-1)) has emerged as a worldwide concern among isolates of Enterobacteriaceae. Its epidemiology is been strongly associated with travel and healthcare on the Indian Subcontinent. We report two cases of urinary tract infection with Enterobacteriaceae
T Galeano Díaz et al.
Journal of chromatography. A, 764(2), 243-248 (1997-03-14)
A HPLC method with coulometric detection has been established to carry out the separation of the three nitrofuran derivatives, nitrofurantoin, furazolidone and furaltadone. A Nova-Pak C18 column (150 x 3.9 mm) and a Coulochem II detector from ESA have been
Thomas Tängdén et al.
Antimicrobial agents and chemotherapy, 54(9), 3564-3568 (2010-06-16)
Foreign travel has been suggested to be a risk factor for the acquisition of extended-spectrum beta-lactamase (ESBL)-producing Enterobacteriaceae. To our knowledge, this has not previously been demonstrated in a prospective study. Healthy volunteers traveling outside Northern Europe were enrolled. Rectal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service